Skip to Content
Merck
All Photos(3)

Key Documents

C2764

Sigma-Aldrich

Activated charcoal

greener alternative

untreated, granular, ≤5 mm

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

<0.1 mmHg ( 20 °C)

Quality Level

form

untreated, granular

autoignition temp.

842 °F

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

1375 μΩ-cm, 20°C (graphite)

particle size

≤5 mm

mp

3550 °C (lit.)

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Activated charcoal (AC) untreated, granular, ≤5 mm is a carbonaceous material that is majorly used in a variety of applications due to its abundance, cost effectiveness and environmentally safe nature. It can be used in the fabrication of energy devices.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

AC can be used as an electrode material in the formation of supercapacitors, double layered capacitors, lithium ion batteries and fuel cells.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harry Marsh
Activated Carbon, Harry Marsh null
Fabrication of textile fabric counter electrodes using activated charcoal doped multi walled carbon nanotube hybrids for dye sensitized solar cells
Arbab AA, et al.
Journal of Material Chemistry A, 4(4), 1495-1505 (2016)
Performance studies of activated charcoal based electrical double layer capacitors with ionic liquid gel polymer electrolytes
Pandey GP, et al.
Energy & Fuels, 24(12), 6644-6652 (2010)
Proton transfer reaction-mass spectrometry as a rapid inline tool for filter efficiency of activated charcoal in support of the development of Solid Oxide Fuel Cells fueled with biogas
Papurello D, et al.
Fuel Processing Technology, 130(12), 78-86 (2015)
Supercapacitors from activated carbon derived from banana fibers
Subramanian V, et al.
The Journal of Physical Chemistry C, 111(20), 7527-7531 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service