Merck
  • A direct PCR approach to accelerate analyses of human-associated microbial communities.
A direct PCR approach to accelerate analyses of human-associated microbial communities.
PloS one (2012-09-11)
Gilberto E Flores, Jessica B Henley, Noah Fierer
ABSTRACT
Since the composition of the human microbiome is highly variable both within and between individuals, researchers are increasingly reliant on high-throughput molecular approaches to identify linkages between the composition of these communities and human health. While new sequencing technologies have made it increasingly feasible to analyze large numbers of human-associated samples, the extraction of DNA from samples often remains a bottleneck in the process. Here we tested a direct PCR approach using the Extract-N-Amp Plant PCR Kit to accelerate the 16S rRNA gene-based analyses of human-associated bacterial communities, directly comparing this method to a more commonly-used approach whereby DNA is first extracted and purified from samples using a series of steps prior to PCR amplification. We used both approaches on replicate samples collected from each of five body habitats (tongue surface, feces, forehead skin, underarm skin, and forearm skin) from four individuals. With the exception of the tongue samples, there were few significant differences in the estimates of taxon richness or phylogenetic diversity obtained using the two approaches. Perhaps more importantly, there were no significant differences between the methods in their ability resolve body habitat differences or inter-individual differences in bacterial community composition and the estimates of the relative abundances of individual taxa were nearly identical with the two methods. Overall, the two methods gave very similar results and the direct PCR approach is clearly advantageous for many studies exploring the diversity and composition of human-associated bacterial communities given that large numbers of samples can be processed far more quickly and efficiently.
Sigma-Aldrich
Extraction Solution, Molecular Biology
Sigma-Aldrich
Extract-N-Amp PCR ReadyMix, Amplifications to support Extract-N-Amp Plant and Extract-N-Amp Tissue
Sigma-Aldrich
Extract-N-Amp Plant PCR Kit, sufficient for 100 extractions, sufficient for 500 amplifications
Sigma-Aldrich
Extract-N-Amp Plant PCR Kit, sufficient for 1000 extractions, sufficient for 1000 amplifications
Sigma-Aldrich
Extract-N-Amp Plant PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications
Sigma-Aldrich
Dilution Solution, Extract-N-Amp Plant Dilution Solution