Merck
  • Reduced postoperative chronic pain after tension-free inguinal hernia repair using absorbable sutures: a single-blind randomized clinical trial.
Reduced postoperative chronic pain after tension-free inguinal hernia repair using absorbable sutures: a single-blind randomized clinical trial.
Journal of the American College of Surgeons (2013-11-12)
Igor Jeroukhimov, Itay Wiser, Evgeny Karasic, Vladimir Nesterenko, Natan Poluksht, Ron Lavy, Ariel Halevy
ABSTRACT
Chronic pain after inguinal hernia repair occurs in 16% to 62% of patients. The underlying mechanism probably involves sensory nerve damage and abnormal healing that might be influenced by the materials chosen for the procedure. We hypothesize that nonabsorbable sutures used for mesh fixation to the surrounding tissues are associated with higher rates of chronic groin pain after surgery. We conducted a single-blind randomized clinical trial to compare the effect of absorbable braided sutures (Vycril; Ethicon) and nonabsorbable monofilament sutures (Prolene; Ethicon) used in inguinal hernia repair on the rate of chronic pain. We assessed chronic pain using a 4-point verbal-rank scale during a 1-year postoperative follow-up period. Study groups included 100 patients in each group. No age, sex, or hernia-side differences were observed between the study groups. Chronic pain rate after surgery was higher in the nonabsorbable monofilament suture group compared with the absorbable braided suture group (37 vs 26 patients; p = 0.056). Time to pain disappearance was longer in the nonabsorbable monofilament suture vs the absorbable braided suture group (115.3 days; 95% CI, 88-142.7 vs 77.4 days; 95% CI, 54.3-100.3; p = 0.038, respectively). A 1-year age increment reduces the risk for chronic pain occurrence by 2.2% (odds ratio = 0.978%; 95% CI, 0.961-0.995; p = 0.013). The use of nonabsorbable sutures increases the risk for chronic pain in 94.9% compared with absorbable sutures (odds ratio = 1.949; 95% CI, 1.039-3.658; p = 0.038). Nonabsorbable suture use in inguinal hernia repair is associated with a higher rate of chronic pain and a longer time to pain disappearance as compared with absorbable sutures.
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, viscosity 0.16-0.24 dL/g 
Sigma-Aldrich
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
Sigma-Aldrich
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
Sigma-Aldrich
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
Sigma-Aldrich
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
Sigma-Aldrich
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000