Merck
  • Comparison of polyvinyl alcohol coated nasal packing with non-absorbable nasal packing.
Comparison of polyvinyl alcohol coated nasal packing with non-absorbable nasal packing.
Rhinology (2013-05-15)
J H Seol, J M Kim, S S Kim, B R Na, H Jung, J H Cho, J K Kim
ABSTRACT
A number of different nasal packing materials are available for prevention of nasal bleeding after endoscopic sinus surgery. Polyvinyl alcohol (PVA) coated nasal packing is an expandable packing for use in rhinological surgery. This innovative surf- ace treatment helps to reduce the possibility of adherence to tissue and of blood clotting within the sponge. The present study investigated the effects of PVA coated packing and non-absorbable packing with respect to pain, healing site and postoperative bleeding following endoscopic sinus surgery. Patients between 18-80 years of age undergoing sinus surgery were enrolled. Each patient`s ethmoid cavities were randomised to receive PVA coated packing material or the standard non-absorbable sinus packs. The remaining nasal packing material was removed on the 2nd day in the clinic. We determined visual analog scale score, bleeding time and wound healing status. A single rhinologist graded postoperative endoscopic appearance. Length of follow-up was 3 months. Thirty three patients were recruited. There was a significant difference in the bleeding time between the two groups, but pain and wound healing were not significantly different between the two groups. PVA-coated nasal packing presents comparable characteristics with traditional nasal packing.
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000