10852
Mowiol® 10-98
Mw ~61,000
Synonym(s):
Poly(vinyl alcohol)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23
Recommended Products
form
solid
Quality Level
mol wt
Mw ~61,000
extent of labeling
98.0-98.8 mol% hydrolysis
~1,400 polymerization
ign. residue
≤0.5%
viscosity
9-11 mPa.s, 4 % in H2O(20 °C)
InChI
1S/C2H4O/c1-2-3/h2-3H,1H2
InChI key
IMROMDMJAWUWLK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Mowiol is a registered trademark of Kuraray Europe GmbH
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
No data available
Flash Point(C)
No data available
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Laura Ramalho et al.
Journal of materials science. Materials in medicine, 31(12), 129-129 (2020-12-01)
In this paper, we created a dynamic adhesive environment (DAE) for adipose tissue-derived mesenchymal stem cells (ADMSCs) cultured on smart thermo-responsive substrates, i.e., poly (N-isopropyl acrylamide) (PNIPAM), via introducing periodic changes in the culture temperature. We further explored the particular
Bor-Sen Chiou et al.
International journal of biological macromolecules, 55, 214-220 (2013-01-29)
Pollock gelatin/poly(vinyl alcohol) (PVA) fibers were electrospun using deionized water as the solvent and pollock gelatin/poly(lactic acid) (PLA) fibers were electrospun using 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) as the solvent. The chemical, thermal, and thermal stability properties were examined for the electrospun samples.
Roger F Falkenstern-Ge et al.
Journal of bronchology & interventional pulmonology, 20(1), 58-62 (2013-01-19)
Emphysema is a progressive and irreversible disease for which there is no cure to date. The patients experience debilitating shortness of breath with repetitive exacerbations and poor quality of daily life. At present, patients with severe emphysema have limited treatment
Miao Zhang et al.
Journal of visualized experiments : JoVE, (73)(73), e4335-e4335 (2013-03-15)
Existing protocols for the generation of neurons from human pluripotent stem cells (hPSCs) are often tedious in that they are multistep procedures involving the isolation and expansion of neural precursor cells, prior to terminal differentiation. In comparison to these time-consuming
Sreejith Raveendran et al.
Carbohydrate polymers, 92(2), 1225-1233 (2013-02-13)
Extremophilic bacterial polysaccharide based biocompatible nanofibers were produced for the first time via electrospinning technique. Mauran (MR), an extremophilic sulfated exopolysaccharide was extracted from moderately halophilic bacterium, Halomonas maura and characterized for the application of nanofiber synthesis. Thin-uniform MR nanofibers
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service