Skip to Content
Merck
All Photos(2)

Documents

363065

Sigma-Aldrich

Poly(vinyl alcohol)

Mw 146,000-186,000, 99+% hydrolyzed

Synonym(s):

PAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23

Quality Level

form

crystalline powder
crystals

mol wt

Mw 146,000-186,000

falling ball

55-65 cP, 4 % in H2O(20 °C)(lit.)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(vinyl alcohol) is a vinyl based water soluble polymer that can be used as a non-ionic surface active agent. It can also be used as a biodegradable polymer and can be incorporated in adhesives, coatings, textiles ceramics and cosmetics. Its properties include good transparency, chemical resistance, toughness and antielectrostatic properties.

Application

PVA can be used as a chelating agent in the synthesis of zirconium oxide nanopowder from zirconium hydroxide. It can also be used as biopolymer which finds potential application in fiber based food packaging.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plastics Materials, 386-397 (1999)
Compostable Polymer Properties and Packaging Applications
Plastic Films in Food Packaging, 217-248 (2013)
Multilayer structures based on annealed electrospun biopolymer coatings of interest in water and aroma barrier fiber-based food packaging applications
Cherpinski A, et al.
Journal of Applied Polymer Science, 135(24), 45501-45501 (2018)
Novel method for synthesis of metastable tetragonal zirconia nanopowders at low temperatures
Kazemi F, et al.
Ceramics-Silikaty, 55(1), 26-30 (2011)
Muhammad Suhaeri et al.
Theranostics, 8(18), 5025-5038 (2018-11-16)
Skin injuries are frequently encountered in daily life, but deep wounds often poorly self-heal and do not recover completely. In this study, we propose a novel skin patch that combines antibiotic, cell-derived extracellular matrix (ECM) and biocompatible polyvinyl alcohol (PVA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service