Merck
  • Oxidative damage of rat liver mitochondria during exposure to t-butyl hydroperoxide. Role of Ca²⁺ ions in oxidative processes.
Oxidative damage of rat liver mitochondria during exposure to t-butyl hydroperoxide. Role of Ca²⁺ ions in oxidative processes.
Life sciences (2013-05-07)
Ilya B Zavodnik, Iosif K Dremza, Vitali T Cheshchevik, Elena A Lapshina, Maria Zamaraewa
ABSTRACT
The present study was designed for further evaluation of the biochemical mechanism of hepatic mitochondrial dysfunction under oxidative damages induced by organic hydroperoxide, tert-butyl hydroperoxide (tBHP), for estimation of the molecular targets impaired during oxidative stress, and for investigation of the role of Ca(2+) ions in mitochondrial oxidative reactions and of the protective effect of melatonin during mitochondrial peroxidative damage. Mitochondria were isolated by differential centrifugation from the rat liver. The effects of tBHP exposure, EDTA, Ca(2+) ions and melatonin on mitochondrial respiratory activity, mitochondrial enzyme activities and redox status were measured. The present study provides evidence that tBHP (at low concentrations of 0.02-0.065mM, in EDTA-free medium) induced uncoupling of the oxidation and phosphorylation processes and decreased the efficiency of the phosphorylation reaction. This effect depended on the respiratory substrate used. The presence of EDTA prevented oxidative impairment of mitochondrial respiration, but Ca(2+) ions in the medium enhanced oxidant-induced mitochondrial damage considerably. In the presence of 0.5mM EDTA, tBHP (at high concentrations, 0.5-2mM) considerably oxidized mitochondrial reduced glutathione, enhanced accumulation of membrane lipid peroxidation products and mixed protein-glutathione disulfides and led to an inhibition of oxoglutarate dehydrogenase and succinate dehydrogenase. Direct oxidative modification of enzymatic complexes of the respiratory chain and mitochondrial matrix, mitochondrial reduced glutathione depletion, protein glutathionylation, membrane lipid peroxidation and Ca(2+) overload are the main events of mitochondrial peroxidative damages. Experiments in vitro demonstrated that melatonin inhibited the mitochondrial peroxidative damage, preventing redox-balance changes and succinate dehydrogenase inactivation.
Product Description
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, puriss. p.a., ≥98.0% (NT)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
Succinic acid disodium salt, 99%
Sigma-Aldrich
Sodium succinate dibasic, purum, anhydrous, ≥98.0% (NT)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N