Skip to Content
Merck
All Photos(4)

Key Documents

E1644

Sigma-Aldrich

Ethylenediaminetetraacetic acid disodium salt dihydrate

98.5-101.5%, BioUltra

Synonym(s):

Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Beilstein:
3900609
EC Number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

product line

BioUltra

Assay

98.5-101.5%

form

powder

reaction suitability

reagent type: chelator

impurities

≤0.005% Phosphorus (P)
≤0.05% NH3
≤0.1% Insoluble matter

pH

4.0-6.0 (0.1 M in H2O)

mp

248 °C (dec.) (lit.)

solubility

H2O: 0.1 M, clear, colorless

cation traces

Al: ≤0.001%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

room temp

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediaminetetraacetic acid (EDTA) is a substituted diamine[1] and a chelator of metal ions.[2] It has anticoagulant[3] and antibacterial activity.[1] EDTA is an inhibitor of metalloprotease.[2] It evades the undesirable effects of ferric, cupric and manganic ions in bleaching. It blocks cellular division, chlorophyll synthesis and algal biomass production.[1]

Application

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modulation of matrix metalloproteinase activity by EDTA prevents posterior capsular opacification
Hazra S, et al.
Molecular Vision, 18, 1701-1701 (2012)
Accurate Results in the Clinical Laboratory: A Guide to Error Detection and Correction (2013)
Steven G Chaulk et al.
Nature protocols, 2(5), 1052-1058 (2007-06-05)
This protocol describes a general method for the preparation of RNAs in which the reactivity or hydrogen-bonding properties of the molecule are modified in a photoreversible fashion by use of a caging strategy. A single caged adenosine, modified at the
Matthew R Miller et al.
Journal of biological methods, 2(3), e23-e23 (2015-10-13)
Over the past two decades, researchers have struggled to efficiently express foreign DNA in primary macrophages, impeding research progress. The applications of lipofection, electroporation, microinjection, and viral-mediated transfer typically result in disruptions in macrophage differentiation and function, low expression levels
Meritxell Espino Guarch et al.
eLife, 7 (2018-01-23)
Age-related hearing loss (ARHL) is the most common sensory deficit in the elderly. The disease has a multifactorial etiology with both environmental and genetic factors involved being largely unknown. SLC7A8/SLC3A2 heterodimer is a neutral amino acid exchanger. Here, we demonstrated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service