Merck
  • Overexpression of O-methyltransferase leads to improved vanillin production in baker's yeast only when complemented with model-guided network engineering.
Overexpression of O-methyltransferase leads to improved vanillin production in baker's yeast only when complemented with model-guided network engineering.
Biotechnology and bioengineering (2012-09-26)
Ana Rita Brochado, Kiran R Patil
ABSTRACT
Overproduction of a desired metabolite is often achieved via manipulation of the pathway directly leading to the product or through engineering of distant nodes within the metabolic network. Empirical examples illustrating the combined effect of these local and global strategies have been so far limited in eukaryotic systems. In this study, we compared the effects of overexpressing a key gene in de novo vanillin biosynthesis (coding for O-methyltransferase, hsOMT) in two yeast strains, with and without model-guided global network modifications. Overexpression of hsOMT resulted in increased vanillin production only in the strain with model-guided modifications, exemplifying advantage of using a global strategy prior to local pathway manipulation.
Product Description
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Vanillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.