Skip to Content
Merck
All Photos(3)

Documents

W310727

Sigma-Aldrich

Vanillin

greener alternative

natural, ≥97%, FCC, FG

Synonym(s):

4-Hydroxy-3-methoxybenzaldehyde, Vanillic aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H3-3-(OCH3)CHO
CAS Number:
Molecular Weight:
152.15
FEMA Number:
3107
Beilstein:
472792
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.018
NACRES:
NA.21

grade

FG
Halal
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 182.60
FDA 21 CFR 182.90

vapor density

5.3 (vs air)

vapor pressure

>0.01 mmHg ( 25 °C)

Assay

≥97%

form

powder

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

170 °C/15 mmHg (lit.)

mp

81-83 °C (lit.)

solubility

water: slightly soluble 10 g/L at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

creamy; sweet; vanilla

SMILES string

COc1cc(C=O)ccc1O

InChI

1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3

InChI key

MWOOGOJBHIARFG-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

Vanillin is the main aroma constituent that contributes to the vanilla flavor. It occurs naturally in the vanilla pods.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product is a Biobased product aligning with the principles of "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks" in Green Chemistry.

Other Notes

produced from rice bran ferulic acid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

319.6 - 321.4 °F - closed cup

Flash Point(C)

159.8 - 160.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanillin and related flavor compounds in vanilla extracts made from beans of various global origins
Ranadive AS.
Journal of Agricultural and Food Chemistry, 40(10), 1922-1924 (1992)
Chia-Lin Lee et al.
Journal of natural products, 72(9), 1568-1572 (2009-08-21)
Two new sesquiterpene coumarins, designated 5'-acetoxy-8'-hydroxyumbelliprenin (1) and 10'R-acetoxy-11'-hydroxyumbelliprenin (2), and a new diterpene, 15-hydroxy-6-en-dehydroabietic acid (3), along with 27 known compounds, were isolated from a CHCl(3)-soluble extract of Ferula assa-foetida through bioassay-guided fractionation. The structures of the new metabolites
Ana Rita Brochado et al.
Biotechnology and bioengineering, 110(2), 656-659 (2012-09-26)
Overproduction of a desired metabolite is often achieved via manipulation of the pathway directly leading to the product or through engineering of distant nodes within the metabolic network. Empirical examples illustrating the combined effect of these local and global strategies
Shixin Deng et al.
Journal of natural products, 69(4), 536-541 (2006-04-29)
Serotonin receptor (5-HT(7)) binding assay-directed fractionation of a methanol extract of the dried roots of Angelica sinensis led to the isolation and identification of 21 compounds including a new phenolic ester, angeliferulate (1), and three new phthalides, 10-angeloylbutylphthalide (2), sinaspirolide
H Priefert et al.
Applied microbiology and biotechnology, 56(3-4), 296-314 (2001-09-11)
Vanillin is one of the most important aromatic flavor compounds used in foods, beverages, perfumes, and pharmaceuticals and is produced on a scale of more than 10 thousand tons per year by the industry through chemical synthesis. Alternative biotechnology-based approaches

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service