Skip to Content
Merck
All Photos(1)

Key Documents

77634

Supelco

Mettler-Toledo Calibration substance ME 51143093, Vanillin

traceable to primary standards (LGC)

Synonym(s):

Vanillin, 4-Hydroxy-3-methoxybenzaldehyde, Vanillic aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H3-3-(OCH3)CHO
CAS Number:
Molecular Weight:
152.15
Beilstein:
472792
EC Number:
MDL number:
UNSPSC Code:
20121904
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.3 (vs air)

vapor pressure

>0.01 mmHg ( 25 °C)

quality

traceable to primary standards (LGC)

shelf life

limited shelf life, expiry date on the label

bp

170 °C/15 mmHg (lit.)

mp

81-83 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

COc1cc(C=O)ccc1O

InChI

1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3

InChI key

MWOOGOJBHIARFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vanillin is a flavor and fragrance compound, found as a major constituent of natural vanilla. It′s of commercial importance in both food and non-food applications and pharmaceutical preparations. It′s well-known as an intermediate in the synthesis of herbicides, antifoaming and antimicrobial agents.

Mettler-Toledo calibration substance ME 51143093, vanillin is an analytical standard for use in the regular checking of Mettler-Toledo melting point instrument. Its value equals an average of 6 to 12 measurements with a Mettler-Toledo MP90 Excellence instrument that is calibrated against primary standards. The melting point is validated by Capillary method according to European Pharmacopeia (2.2.14.)

Application

Vanillin Mettler-Toledo calibration substance ME 51143093 is a melting point calibration standard used to calibrate Mettler-Toledo mp instruments.

Features and Benefits

  • Traceable to a primary standard (LGC, London)
  • Melting point evaluation conducted in both thermodynamic and pharmacopeia modes for physically correct and heating rate dependent melting point determinations, respectively
  • Provided with certificates of analysis and safety data sheet
  • A product of analytical standard grade to help meet the QC/QA requirements of melting point determination
  • Standard deviation up to ± 0.2 °C

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

319.6 - 321.4 °F - closed cup

Flash Point(C)

159.8 - 160.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanillin
Walton NJ, et al.
Phytochemistry, 505-515 null
Ana Rita Brochado et al.
Biotechnology and bioengineering, 110(2), 656-659 (2012-09-26)
Overproduction of a desired metabolite is often achieved via manipulation of the pathway directly leading to the product or through engineering of distant nodes within the metabolic network. Empirical examples illustrating the combined effect of these local and global strategies
Igor A O Reis et al.
Food chemistry, 135(4), 2453-2461 (2012-09-18)
A single-step selective separation of two food additives was investigated using alcohol-salt aqueous two-phase systems (ATPS). The selective partitioning of two of the most used additives from a processed food waste material, vanillin and l-ascorbic acid, was successfully accomplished. The
Yavuz Yardım et al.
Food chemistry, 141(3), 1821-1827 (2013-07-23)
A method for the determination of food additive vanillin was developed by adsorptive stripping voltammetry. Its determination was carried out at the anodically pre-treated boron-doped diamond electrode in aqueous solutions. Using square-wave stripping mode, the compound yielded a well-defined voltammetric
Sartaj Tabassum et al.
European journal of medicinal chemistry, 60, 216-232 (2013-01-08)
SOD mimics with varying coligand are momentous in developing potential chemotherapeutic drugs. Cu(II) based SOD mimics 1-4 [CuLH(OAc)(H(2)O)Y)] (LH = 2-((E)-(1,3-dihydroxy-2-methylpropan-2-ylimino)methyl)-6-methoxyphenol, OAc = CH(3)COO, 1: Y = H(2)O; 2: Y = phen (1,10-phenanthroline), 3: Y = tpimH (2,4,5-triphenylimidazole); 4: Y

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service