Merck
Microglia regulate central nervous system myelin growth and integrity.
Nature (2022-12-15)
Niamh B McNamara, David A D Munro, Nadine Bestard-Cuche, Akiko Uyeda, Jeroen F J Bogie, Alana Hoffmann, Rebecca K Holloway, Irene Molina-Gonzalez, Katharine E Askew, Stephen Mitchell, William Mungall, Michael Dodds, Carsten Dittmayer, Jonathan Moss, Jamie Rose, Stefan Szymkowiak, Lukas Amann, Barry W McColl, Marco Prinz, Tara L Spires-Jones, Werner Stenzel, Karen Horsburgh, Jerome J A Hendriks, Clare Pridans, Rieko Muramatsu, Anna Williams, Josef Priller, Veronique E Miron
ABSTRACT
Myelin is required for the function of neuronal axons in the central nervous system, but the mechanisms that support myelin health are unclear. Although macrophages in the central nervous system have been implicated in myelin health1, it is unknown which macrophage populations are involved and which aspects they influence. Here we show that resident microglia are crucial for the maintenance of myelin health in adulthood in both mice and humans. We demonstrate that microglia are dispensable for developmental myelin ensheathment. However, they are required for subsequent regulation of myelin growth and associated cognitive function, and for preservation of myelin integrity by preventing its degeneration. We show that loss of myelin health due to the absence of microglia is associated with the appearance of a myelinating oligodendrocyte state with altered lipid metabolism. Moreover, this mechanism is regulated through disruption of the TGFβ1-TGFβR1 axis. Our findings highlight microglia as promising therapeutic targets for conditions in which myelin growth and integrity are dysregulated, such as in ageing and neurodegenerative disease2,3.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody, clone 8-18C5, Chemicon®, from mouse
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-CNP antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2887, purified immunoglobulin, buffered aqueous glycerol solution
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-Myelin Associated Glycoprotein Antibody, clone 513, clone 513, Chemicon®, from mouse
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse