Skip to Content
Merck
All Photos(1)

Documents

MAB5680

Sigma-Aldrich

Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody

clone 8-18C5, Chemicon®, from mouse

Synonym(s):

MOG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8-18C5, monoclonal

species reactivity

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

GenBank accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Mog(17441)

General description

Myelin oligodendrocyte glycoprotein (MOG) is a key CNS-specific autoantigen for primary demyelination in multiple sclerosis. Although the disease-inducing role of MOG has been established, its precise function in the CNS remains obscure. MOG is a type I integral membrane protein possessing a single extracellular Ig variable domain (Ig-V) (3, 13, 14). The amino acid sequence of MOG is highly conserved among animal species (>90%), indicative of an important biological function. MOG is specifically expressed in the CNS on the outermost lamellae of the myelin sheath as well as the cell body and processes of oligodendrocytes. The developmentally late expression of MOG correlates with the later stages of myelinogenesis, suggesting that MOG has a role in the completion, compaction, and/or maintenance of myelin, further suggesting that MOG has an adhesive function within the CNS . Consistent with MOG′s possible adhesive role in the CNS, a homodimeric form of MOG has not only been observed after isolation from the CNS but has additionally been observed in situ.

Specificity

May cross react with human and bovine based on sequence similarity.
Membane associated region of MOG [Myelin oligodendrocyte glycoprotein].

Immunogen

Epitope: The antibody binds to a discontinuous epitope present on the extracellular immunoglobulin V-like domain of MOG.
Rat cerebellar glycoproteins

Application

Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody is an antibody against Myelin Oligodendrocyte Glycoprotein (MOG) for use in FC, IC, IH & WB.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Quality

Routinely evaluated by immunoblot.

Target description

28 kDa

Physical form

Format: Purified
Purified immunoglobulin
Purified in 0.1 M Tris-Glycine, 0.15 M NaCl, pH 7.4 and 0.05% Sodium Azide

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 1 year after date of receipt.

Analysis Note

Control
brain lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Xu et al.
The Journal of experimental medicine, 215(4), 1169-1186 (2018-03-01)
The pathophysiology of drug-resistant pediatric epilepsy is unknown. Flow cytometric analysis of inflammatory leukocytes in resected brain tissues from 29 pediatric patients with genetic (focal cortical dysplasia) or acquired (encephalomalacia) epilepsy demonstrated significant brain infiltration of blood-borne inflammatory myeloid cells
Alvaro Cobo-Calvo et al.
Neurology(R) neuroimmunology & neuroinflammation, 6(2), e543-e543 (2019-02-26)
To describe clinical and radiologic features of cranial nerve (CN) involvement in patients with myelin oligodendrocyte glycoprotein antibodies (MOG-IgG) and to assess the potential underlying mechanism of CN involvement using a nonhuman primate (NHP) model. Epidemiologic, clinical, and radiologic features
Fadi J Najm et al.
Nature biotechnology, 31(5), 426-433 (2013-04-16)
Cell-based therapies for myelin disorders, such as multiple sclerosis and leukodystrophies, require technologies to generate functional oligodendrocyte progenitor cells. Here we describe direct conversion of mouse embryonic and lung fibroblasts to induced oligodendrocyte progenitor cells (iOPCs) using sets of either
Rong He et al.
Molecular medicine reports, 16(1), 119-126 (2017-05-24)
Fasudil has been demonstrated to possess a protective effect in neural injury; however, its protective effect on convulsive brain injury remains to be assessed. The aim of the present study was to investigate the latent mechanism and effect of fasudil
Zhaohui Shao et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(12), 3127-3137 (2017-02-15)
Differentiation and maturation of oligodendrocyte progenitor cells (OPCs) involve the assembly and disassembly of actin microfilaments. However, how actin dynamics are regulated during this process remains poorly understood. Leucine-rich repeat and Ig-like domain-containing Nogo receptor interacting protein 1 (LINGO-1) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service