Merck
  • Visible light responsive photocatalyst induces progressive and apical-terminus preferential damages on Escherichia coli surfaces.
Visible light responsive photocatalyst induces progressive and apical-terminus preferential damages on Escherichia coli surfaces.
PloS one (2011-05-19)
Je-Wen Liou, Ming-Hui Gu, Yen-Kai Chen, Wen-Yi Chen, Yi-Cheng Chen, Yao-Hsuan Tseng, Yu-Jiun Hung, Hsin-Hou Chang
ABSTRACT
Recent research shows that visible-light responsive photocatalysts have potential usage in antimicrobial applications. However, the dynamic changes in the damage to photocatalyzed bacteria remain unclear. Facilitated by atomic force microscopy, this study analyzes the visible-light driven photocatalyst-mediated damage of Escherichia coli. Results show that antibacterial properties are associated with the appearance of hole-like structures on the bacteria surfaces. Unexpectedly, these hole-like structures were preferentially induced at the apical terminus of rod shaped E. coli cells. Differentiating the damages into various levels and analyzing the percentage of damage to the cells showed that photocatalysis was likely to elicit sequential damages in E. coli cells. The process began with changing the surface properties on bacterial cells, as indicated in surface roughness measurements using atomic force microscopy, and holes then formed at the apical terminus of the cells. The holes were then subsequently enlarged until the cells were totally transformed into a flattened shape. Parallel experiments indicated that photocatalysis-induced bacterial protein leakage is associated with the progression of hole-like damages, further suggesting pore formation. Control experiments using ultraviolet light responsive titanium-dioxide substrates also obtained similar observations, suggesting that this is a general phenomenon of E. coli in response to photocatalysis. The photocatalysis-mediated localization-preferential damage to E. coli cells reveals the weak points of the bacteria. This might facilitate the investigation of antibacterial mechanism of the photocatalysis.
Product Description
Latex beads, polystyrene, 0.1 μm mean particle size
Latex beads, polystyrene, 1.1 μm mean particle size
Latex beads, polystyrene, 0.8 μm mean particle size
Latex beads, polystyrene, 0.46 μm mean particle size
Latex beads, polystyrene, 3.0 μm mean particle size
Latex beads, polystyrene, 0.3 μm mean particle size
Latex beads, polystyrene, 0.6 μm mean particle size