Skip to Content
Merck

Skip To

LB5

Latex beads, polystyrene

0.46 μm mean particle size

Synonym(s):

Latex microspheres, Polystyrene beads, Polystyrene microspheres, plastic beads

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous suspension

composition

Solids, 10%

packaging

pack of 1 mL, pack of 15 mL, pack of 2 mL

mean particle size

0.46 μm

General description

Polystyrene microparticles are negativelycharged- stabilized colloidal particles. These microparticles are synthesizedby the polymerization of styrene under conditions that produce coalescent beadformation. Polystyrene latex beads have a broad range of applications such as cellcounter calibration, antibody-mediated agglutination diagnostics, electronmicroscopy, and phagocytosis experiments.

Application

Polystyrene latex beads have been used as a component of an artificial device to draw blood from mice. It has also been used as a component of the protein-free latex meal for the extraction of the peritrophic matrix (PM) from female Anopheles gambiae.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
LB1LB3LB8
form

aqueous suspension

form

aqueous suspension

form

aqueous suspension

form

aqueous suspension

mean particle size

0.46 μm

mean particle size

0.1 μm

mean particle size

0.3 μm

mean particle size

0.8 μm

packaging

pack of 1 mL, pack of 2 mL

packaging

pack of 1 ea

packaging

pack of 1 mL, pack of 15 mL, pack of 2 mL

packaging

pack of 1 mL, pack of 15 mL, pack of 2 mL

composition

Solids, 10%

composition

Solids, 10%

composition

Solids, 10%

composition

Solids, 10%


Still not finding the right product?

Explore all of our products under Latex beads, polystyrene


hcodes

pcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library