Skip to Content
Merck
All Photos(1)

Documents

LB3

Latex beads, polystyrene

0.3 μm mean particle size

Synonym(s):

Latex Microspheres, Polystyrene Beads, Polystyrene Latex Particles

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41121800
NACRES:
NB.22

form

aqueous suspension

composition

Solids, 10%

packaging

pack of 1 mL
pack of 15 mL
pack of 2 mL

mean particle size

0.3 μm

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene microparticles are stabilized colloidal particles that are anionic. They are producedby the polymerization of styrene under conditions that induce spontaneouscoalescent bead formation. Latex particles are ideal for phagocytosisexperiments, antibody-mediated agglutination diagnostics, electron microscopy,and cell counter calibration.

Application

Polystyrene latex beads have been used:
  • in latex particle recovery and size distribution studies
  • as a component of the colloidal suspensions for light extinction spectroscopy
  • in the quantification of microvesicles by high-sensitivity flow cytometry

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Chen et al.
Journal of biophotonics, 10(11), 1411-1420 (2017-04-19)
Optical coherence microscopy (OCM) has unique advantages of non-invasive 3D imaging without the need of exogenous labels for studying biological samples. However, the imaging depth of this technique is limited by the tradeoff between the depth of focus (DOF) and
Light extinction spectroscopy applied to polystyrene colloids: Sensitivity to complex refractive index uncertainties and to noise
Eneren P, et al.
Journal of Quantitative Spectroscopy & Radiative Transfer, 261, 107494-107494 (2021)
Richard Ortega et al.
Particle and fibre toxicology, 11, 14-14 (2014-03-29)
The mechanisms of toxicity of metal oxide particles towards lung cells are far from being understood. In particular, the relative contribution of intracellular particulate versus solubilized fractions is rarely considered as it is very challenging to assess, especially for low-solubility
Bacterial cell recovery after hollow fiber microfiltration sample concentration: Most probable bacterial composition in frozen vegetables
Irwin P, et al.
LWT--Food Science and Technology, 140, 110647-110647 (2021)
Hemofiltration induces generation of leukocyte-derived CD31+/CD41? microvesicles in sepsis
Georg F, et al.
Annals of intensive care (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service