Merck
  • Structure-activity relationships of trans-substituted-propenoic acid derivatives on the nicotinic acid receptor HCA2 (GPR109A).
Structure-activity relationships of trans-substituted-propenoic acid derivatives on the nicotinic acid receptor HCA2 (GPR109A).
Bioorganic & medicinal chemistry letters (2010-12-21)
J P D van Veldhoven, C C Blad, C M Artsen, C Klopman, D R Wolfram, M J Abdelkadir, J R Lane, J Brussee, A P Ijzerman
ABSTRACT
Nicotinic acid (niacin) has been used for decades as an antidyslipidemic drug in man. Its main target is the hydroxy-carboxylic acid receptor HCA2 (GPR109A), a G protein-coupled receptor. Other acids and esters such as methyl fumarate also interact with the receptor, which constituted the basis for the current study. We synthesized a novel series of substituted propenoic acids, such as fumaric acid esters, fumaric acid amides and cinnamic acid derivatives, and determined their affinities for the HCA2 receptor. We observed a rather restricted binding pocket on the receptor with trans-cinnamic acid being the largest planar ligand in our series with appreciable affinity for the receptor. Molecular modeling and analysis of the structure-activity relationships in the series suggest a planar trans-propenoic acid pharmacophore with a maximum length of 8 Å and out-of-plane orientation of the larger substituents.
Sigma-Aldrich
2-Pentenoic acid, predominantly trans, 98%
Sigma-Aldrich
3,3-Dimethylacrylic acid, 97%
Sigma-Aldrich
trans-3-Hydroxycinnamic acid, 99%
Sigma-Aldrich
4-Methylcinnamic acid, predominantly trans, 99%
Sigma-Aldrich
trans-3-(3-Pyridyl)acrylic acid, 99%
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
mono-Ethyl fumarate, 95%
Sigma-Aldrich
trans-Cinnamic acid, 97%
Sigma-Aldrich
Fumaric acid, ≥99.0% (T)
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Crotonic acid, 98%
Sigma-Aldrich
Fumaric acid, FCC, FG
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
mono-Methyl fumarate, 97%