Skip to Content
Merck
All Photos(1)

Key Documents

W228818

Sigma-Aldrich

trans-Cinnamic acid

≥99%, FG

Synonym(s):

trans-3-Phenylacrylic acid, Cinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOOH
CAS Number:
Molecular Weight:
148.16
FEMA Number:
2288
Beilstein:
1905952
EC Number:
Council of Europe no.:
22
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.022
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥99%

bp

300 °C (lit.)

mp

132-135 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cinnamon; honey; floral; spicy; sweet

SMILES string

OC(=O)\C=C\c1ccccc1

InChI

1S/C9H8O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H,(H,10,11)/b7-6+

InChI key

WBYWAXJHAXSJNI-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Therapeutic Implications of Phenolic Acids for Ameliorating Inflammatory Bowel Disease.: This review highlights the therapeutic potential of phenolic acids, including trans-cinnamic acid, in treating inflammatory bowel disease (IBD). The anti-inflammatory properties of trans-cinnamic acid are discussed, showcasing its promise as a natural treatment option for IBD (Lu and Han, 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bianca C Pérez et al.
Journal of medicinal chemistry, 56(2), 556-567 (2013-01-01)
The control of malaria is challenged by drug resistance, and new antimalarial drugs are needed. New drug discovery efforts include consideration of hybrid compounds as potential multitarget antimalarials. Previous work from our group has demonstrated that hybrid structures resulting from
Marie S Prevost et al.
Journal of medicinal chemistry, 56(11), 4619-4630 (2013-05-21)
Pentameric ligand gated ion channels (pLGICs) mediate signal transduction. The binding of an extracellular ligand is coupled to the transmembrane channel opening. So far, all known agonists bind at the interface between subunits in a topologically conserved "orthosteric site" whose
Ying Lei et al.
International journal of pharmaceutics, 453(2), 579-586 (2013-06-13)
Herein, we reported a new type of self-assembly micelles based on amphiphilic polymers of cinnamate and coumarin derivatives modified PEG for drug delivery applications. Lipophilic cinnamic acid (CIN) and 7-carboxyl methoxycoumarin (COU) were immobilized on the terminal groups of poly(ethylene
Qian Fan et al.
Food chemistry, 134(2), 1081-1087 (2012-10-31)
Nine cinnamoyl amides with amino acid ester (CAAE) moiety were synthesized by the conjugation of the corresponding cinnamic acids (cinnamic acid, 4-hydroxy cinnamic acid, ferulic acid and caffeic acid) with amino acid esters, and their inhibitory effects on the activities
Stanislava Gorjanović et al.
Journal of agricultural and food chemistry, 60(38), 9573-9580 (2012-09-07)
Hydrogen peroxide scavenging (HPS) activity of unfermented (green, yellow, and white), partially fermented (oolong), and completely fermented (black) tea ( Camellia sinensis ), maté ( Ilex paraguariensis ), and various herbal infusions, as well as individual compounds (flavan-3-ols, flavonols, cinnamic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service