Skip to Content
Merck
All Photos(2)

Documents

651419

Sigma-Aldrich

mono-Methyl fumarate

97%

Synonym(s):

Fumaric acid monomethyl ester, Methyl hydrogen fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH=CHCO2CH3
CAS Number:
Molecular Weight:
130.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

144-145 °C (lit.)

SMILES string

COC(=O)\C=C\C(O)=O

InChI

1S/C5H6O4/c1-9-5(8)3-2-4(6)7/h2-3H,1H3,(H,6,7)/b3-2+

InChI key

NKHAVTQWNUWKEO-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

mono-Methyl fumarate can be used as a reactant to synthesize:
  • Jumonji C domain-containing histone demethylases (JCHDMs) inhibitor.
  • (−)-Xylariamide A, a fungal metabolite.
  • (±)-Methoxyfumimycin ethyl ester, a potential bacterial peptide deformylase inhibitor.

It is also a wide spectrum antibacterial agent with a powerful antioxidant activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A selective inhibitor and probe of the cellular functions of Jumonji C domain-containing histone demethylases.
Luo X, et al.
Journal of the American Chemical Society, 133(24), 9451-9456 (2011)
A concise synthesis of (?)-methoxyfumimycin ethyl ester.
Zhou Z, et al.
J. Chem. Res. (M), 38(6) (2014)
Jennifer N Hahn et al.
Journal of immunology (Baltimore, Md. : 1950), 197(10), 3850-3860 (2016-10-14)
Extracellular matrix metalloproteinase inducer (EMMPRIN, CD147) is a transmembrane glycoprotein that is upregulated on leukocytes in active lesions in multiple sclerosis (MS) and its animal model, experimental autoimmune encephalomyelitis (EAE). Administration of anti-EMMPRIN Abs reduces the severity of EAE. Minocycline
Laura Northrup et al.
Molecular pharmaceutics, 14(1), 66-80 (2017-01-04)
Current therapies to treat autoimmune diseases often result in side effects such as nonspecific immunosuppression. Therapies that can induce antigen-specific immune tolerance provide an opportunity to reverse autoimmunity and mitigate the risks associated with global immunosuppression. In an effort to
Modification of chitosan with monomethyl fumaric acid in an ionic liquid solution.
Wang Z, et al.
Carbohydrate Polymers, 117, 973-979 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service