Merck
  • Air to lung partition coefficients for volatile organic compounds and blood to lung partition coefficients for volatile organic compounds and drugs.
Air to lung partition coefficients for volatile organic compounds and blood to lung partition coefficients for volatile organic compounds and drugs.
European journal of medicinal chemistry (2007-06-05)
Michael H Abraham, Adam Ibrahim, William E Acree
ABSTRACT
Values of in vitro gas to lung partition coefficients, K(lung), of VOCs have been collected from the literature. For 44 VOCs, application of the Abraham solvation equation to log K(lung) yielded a correlation with R(2)=0.968 and S.D.=0.25 log units. Combination of the log K(lung) values with log K(blood) values leads to in vitro blood to lung partition coefficients, log P(lung) for 43 VOCs; an Abraham solvation equation correlated these values with a very poor R(2)=0.262 but with a very good S.D.=0.190 log units. Values of in vivo log P(lung) for 80 drugs were collected, and were correlated with R(2)=0.647 and S.D.=0.51 log units. When the log P(lung) values for VOCs and drugs were combined, an Abraham solvation equation could correlate the 123 compounds with R(2)=0.676 and S.D.=0.43 log units. Division of the 123 compounds into a training set and a test set, showed that the training equation could predict log P(lung) values with an average error of 0.001 and a standard deviation of 0.44 log units; for drugs in the combined test set the average error was 0.02 and the standard deviation 0.43 log units.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Chloroform solution, NMR reference standard, 3% in acetone-d6 (99.9 atom % D)
Sigma-Aldrich
Chloroform solution, NMR reference standard, 50% in acetone-d6 (99.9 atom % D), chromium(III) acetylacetonate 0.2 %
Supelco
Chloroform solution, certified reference material, 5000 μg/mL in methanol
Supelco
Ethylene oxide solution, certified reference material, 50 mg/mL in methanol
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Pyrene, analytical standard
Supelco
Chloroform solution, NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Supelco
Chloroform solution, NMR reference standard, 20% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Tetrachloroethylene, anhydrous, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Supelco
Digoxin, analytical standard
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder