Merck
  • Repurposing a neurodegenerative disease drug to treat Gram-negative antibiotic-resistant bacterial sepsis.
Repurposing a neurodegenerative disease drug to treat Gram-negative antibiotic-resistant bacterial sepsis.
Science translational medicine (2020-11-20)
David M P De Oliveira, Lisa Bohlmann, Trent Conroy, Freda E-C Jen, Arun Everest-Dass, Karl A Hansford, Raghu Bolisetti, Ibrahim M El-Deeb, Brian M Forde, Minh-Duy Phan, Jake A Lacey, Aimee Tan, Tania Rivera-Hernandez, Stephan Brouwer, Nadia Keller, Timothy J Kidd, Amanda J Cork, Michelle J Bauer, Gregory M Cook, Mark R Davies, Scott A Beatson, David L Paterson, Alastair G McEwan, Jian Li, Mark A Schembri, Mark A T Blaskovich, Michael P Jennings, Christopher A McDevitt, Mark von Itzstein, Mark J Walker
ABSTRACT
The emergence of polymyxin resistance in carbapenem-resistant and extended-spectrum β-lactamase (ESBL)-producing bacteria is a critical threat to human health, and alternative treatment strategies are urgently required. We investigated the ability of the hydroxyquinoline analog ionophore PBT2 to restore antibiotic sensitivity in polymyxin-resistant, ESBL-producing, carbapenem-resistant Gram-negative human pathogens. PBT2 resensitized Klebsiella pneumoniae, Escherichia coli, Acinetobacter baumannii, and Pseudomonas aeruginosa to last-resort polymyxin class antibiotics, including the less toxic next-generation polymyxin derivative FADDI-287, in vitro. We were unable to select for mutants resistant to PBT2 + FADDI-287 in polymyxin-resistant E. coli containing a plasmid-borne mcr-1 gene or K. pneumoniae carrying a chromosomal mgrB mutation. Using a highly invasive K. pneumoniae strain engineered for polymyxin resistance through mgrB mutation, we successfully demonstrated the efficacy of PBT2 + polymyxin (colistin or FADDI-287) for the treatment of Gram-negative sepsis in immunocompetent mice. In comparison to polymyxin alone, the combination of PBT2 + polymyxin improved survival and reduced bacterial dissemination to the lungs and spleen of infected mice. These data present a treatment modality to break antibiotic resistance in high-priority polymyxin-resistant Gram-negative pathogens.
Sigma-Aldrich
Meropenem trihydrate, ≥98% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Gentamicin solution, 10 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amikacin disulfate salt, potency: 674-786 μg per mg (as amikacin base)
Sigma-Aldrich
Doxycycline hydrochloride
Supelco
Anhydrotetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Colistin sulfate salt, ≥19,000 IU/mg
Sigma-Aldrich
Polymyxin B sulfate salt
Sigma-Aldrich
Trimethoprim, ≥98.5%
Supelco
Cefoxitin sodium salt
Sigma-Aldrich
Ceftazidime hydrate