Skip to Content
Merck
All Photos(1)

Key Documents

A1774

Sigma-Aldrich

Amikacin disulfate salt

potency: 674-786 μg per mg (as amikacin base)

Synonym(s):

(2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-(3-amino-3-deoxy-alpha-D-glucopyranosyloxy)-4-(6-amino-6-deoxy-alpha-D-glucopyranosyloxy)-3-hydroxycyclohexyl]-2-hydroxybutanamide disulfate, Antibiotic BB-K8 sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43N5O13 · 2H2SO4
CAS Number:
Molecular Weight:
781.76
Beilstein:
6172633
EC Number:
MDL number:
UNSPSC Code:
51281632
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

biological source

synthetic

form

powder or crystals

potency

674-786 μg per mg (as amikacin base)

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria
mycobacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O

InChI

1S/C22H43N5O13.2H2O4S/c23-2-1-8(29)20(36)27-7-3-6(25)18(39-22-16(34)15(33)13(31)9(4-24)37-22)17(35)19(7)40-21-14(32)11(26)12(30)10(5-28)38-21;2*1-5(2,3)4/h6-19,21-22,28-35H,1-5,23-26H2,(H,27,36);2*(H2,1,2,3,4)/t6-,7+,8-,9+,10+,11-,12+,13+,14+,15-,16+,17-,18+,19-,21+,22+;;/m0../s1

InChI key

FXKSEJFHKVNEFI-GCZBSULCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Amikacin is an aminoglycoside antibiotic commonly used in the treatment of drug-resistant mycobacteria [1]. It is used to study organism-directed delivery of antibiotics as well as drug resistance [2].

Biochem/physiol Actions

Amikacin prevents bacterial protein synthesis by binding to the 30S ribosome subunit and inducing mRNA misreading.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Lovering et al.
The Journal of antimicrobial chemotherapy, 43(5), 719-721 (1999-06-26)
Antibiotic-free human serum was spiked with known concentrations of liposomal amikacin and assayed on the Abbott TDx System, using polarization fluoroimmuno assay (PFIA) kits from Abbott Laboratories, Oxis and Sigma. Although all three kits gave a linear response, the Abbott
Targeted Delivery of Amikacin into Granuloma.
Ana Montes-Worboys, et al.
American Journal of Respiratory Cell and Molecular Biology, 182, 1546-1553 (2010)
Kathryn Y Burge et al.
Journal of human lactation : official journal of International Lactation Consultant Association, 35(3), 538-549 (2019-05-06)
Human milk is known to be protective against necrotizing enterocolitis, a devastating intestinal inflammatory disease affecting the preterm population. Although the pathogenesis of necrotizing enterocolitis is yet to be solidified, intestinal integrity dysfunction, bacterial invasion and/or translocation, and inflammation may
Kenneth S Chen et al.
Pediatric blood & cancer, 60(11), 1772-1777 (2013-06-22)
Children undergoing cancer therapy often receive aminoglycosides to treat febrile neutropenia or gram-negative infections. The magnitude of the risk of developing aminoglycoside-induced ototoxicity and the dose threshold at which that risk significantly increases are unknown. Eligible cancer patients received the
Sabine Ladrech et al.
The European journal of neuroscience, 38(6), 2962-2972 (2013-07-10)
To examine whether an inflammatory process occurs in the amikacin-poisoned cochlea, we investigated the presence of the cytokines tumour necrosis factor-α (TNF-α), interleukin (IL)-1β, and IL-10. No TNF-α, IL-1β or IL-10 was detected in the cochlear perilymph after the loss

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service