Merck
  • Lactate Elicits ER-Mitochondrial Mg2+ Dynamics to Integrate Cellular Metabolism.
Lactate Elicits ER-Mitochondrial Mg2+ Dynamics to Integrate Cellular Metabolism.
Cell (2020-10-10)
Cassidy C Daw, Karthik Ramachandran, Benjamin T Enslow, Soumya Maity, Brian Bursic, Matthew J Novello, Cherubina S Rubannelsonkumar, Ayah H Mashal, Joel Ravichandran, Terry M Bakewell, Weiwei Wang, Kang Li, Travis R Madaris, Christopher E Shannon, Luke Norton, Soundarya Kandala, Jeffrey Caplan, Subramanya Srikantan, Peter B Stathopulos, W Brian Reeves, Muniswamy Madesh
ABSTRACT
Mg2+ is the most abundant divalent cation in metazoans and an essential cofactor for ATP, nucleic acids, and countless metabolic enzymes. To understand how the spatio-temporal dynamics of intracellular Mg2+ (iMg2+) are integrated into cellular signaling, we implemented a comprehensive screen to discover regulators of iMg2+ dynamics. Lactate emerged as an activator of rapid release of Mg2+ from endoplasmic reticulum (ER) stores, which facilitates mitochondrial Mg2+ (mMg2+) uptake in multiple cell types. We demonstrate that this process is remarkably temperature sensitive and mediated through intracellular but not extracellular signals. The ER-mitochondrial Mg2+ dynamics is selectively stimulated by L-lactate. Further, we show that lactate-mediated mMg2+ entry is facilitated by Mrs2, and point mutations in the intermembrane space loop limits mMg2+ uptake. Intriguingly, suppression of mMg2+ surge alleviates inflammation-induced multi-organ failure. Together, these findings reveal that lactate mobilizes iMg2+ and links the mMg2+ transport machinery with major metabolic feedback circuits and mitochondrial bioenergetics.
Sigma-Aldrich
Sodium D-lactate, ≥99.0% (NT)
Sigma-Aldrich
Sodium fumarate dibasic, ≥99%
Sigma-Aldrich
Adenosine 5′-triphosphate magnesium salt, ≥95%, bacterial
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Sigma-Aldrich
3-Phenyllactic acid, ≥98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Octoclothepin maleate salt, solid
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Adenosine 5′-triphosphate dipotassium salt hydrate, from microbial, ≥92% (HPLC)
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Sodium L-lactate, ~98%
Sigma-Aldrich
MISSION® esiRNA, targeting human MCTS1
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Premixed 32 Plex - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ionomycin from Streptomyces conglobatus, ≥98% (HPLC)
Sigma-Aldrich
(+)-Methyl D-lactate, 98%, optical purity ee: 96% (GLC)
Sigma-Aldrich
Williams′ Medium E, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
α-Ketoglutaric acid disodium salt dihydrate, ≥98.0% (dried material, NT)
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
(−)-Methyl L-lactate, 98%, optical purity ee: 97% (GLC)
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
Adenosine 5′-diphosphate sodium salt, bacterial, ≥95% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
L-(+)-Lactic acid, ≥98%