Merck
  • Identification and characterization of novel benzil (diphenylethane-1,2-dione) analogues as inhibitors of mammalian carboxylesterases.
Identification and characterization of novel benzil (diphenylethane-1,2-dione) analogues as inhibitors of mammalian carboxylesterases.
Journal of medicinal chemistry (2005-04-15)
Randy M Wadkins, Janice L Hyatt, Xin Wei, Kyoung Jin P Yoon, Monika Wierdl, Carol C Edwards, Christopher L Morton, John C Obenauer, Komath Damodaran, Paul Beroza, Mary K Danks, Philip M Potter
ABSTRACT
Carboxylesterases (CE) are ubiquitous enzymes responsible for the metabolism of xenobiotics. Because the structural and amino acid homology among esterases of different classes, the identification of selective inhibitors of these proteins has proved problematic. Using Telik's target-related affinity profiling (TRAP) technology, we have identified a class of compounds based on benzil (1,2-diphenylethane-1,2-dione) that are potent CE inhibitors, with K(i) values in the low nanomolar range. Benzil and 30 analogues demonstrated selective inhibition of CEs, with no inhibitory activity toward human acetylcholinesterase or butyrylcholinesterase. Analysis of structurally related compounds indicated that the ethane-1,2-dione moiety was essential for enzyme inhibition and that potency was dependent on the presence of, and substitution within, the benzene ring. 3D-QSAR analyses of these benzil analogues for three different mammalian CEs demonstrated excellent correlations of observed versus predicted K(i) (r(2) > 0.91), with cross-validation coefficients (q(2)) of 0.9. Overall, these results suggest that selective inhibitors of CEs with potential for use in clinical applications can be designed.
Sigma-Aldrich
1,3-Cyclohexanedione, 97%
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Supelco
Benzophenone, analytical standard
Sigma-Aldrich
Hydrobenzoin
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Supelco
Acetylacetone, analytical standard
Sigma-Aldrich
Acetylacetone, Wacker Chemie AG, ≥99.5% (GC)
Sigma-Aldrich
2,2-Dimethoxy-2-phenylacetophenone, 99%
Sigma-Aldrich
Tetrachloro-1,4-benzoquinone, 99%
Sigma-Aldrich
Methyl cyclopentenolone, anhydrous, 98%, FCC, FG
Sigma-Aldrich
Acetylacetone, ReagentPlus®, ≥99%
Sigma-Aldrich
1,3-Diphenyl-1,3-propanedione, 98%
Sigma-Aldrich
4-Benzoylbenzoic acid, 99%
Sigma-Aldrich
Benzoin, purified by sublimation, ≥99.5%
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%
Sigma-Aldrich
2-Methyl-1,3-cyclopentanedione, 99%
Sigma-Aldrich
3-Methyl-1,2-cyclopentanedione, 99%
Sigma-Aldrich
Benzoin, 98%
Supelco
2,3-Butanedione, analytical standard
Supelco
p-Benzoquinone, for spectrophotometric det. of amines, ≥99.5% (HPLC)
Supelco
Chloranil, PESTANAL®, analytical standard
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Sigma-Aldrich
2,3-Butanedione, 97%
Sigma-Aldrich
2-Methyl-1,3-cyclohexanedione, 97%
Sigma-Aldrich
3,4,5,6-Tetrachloro-1,2-benzoquinone, 97%
Sigma-Aldrich
1-Phenyl-1,2-propanedione, 98%, FG
Sigma-Aldrich
4-Methylumbelliferyl acetate, esterase substrate