Skip to Content
Merck
All Photos(1)

Key Documents

05581

Supelco

Acetylacetone

analytical standard

Synonym(s):

2,4-Pentanedione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COCH3
CAS Number:
Molecular Weight:
100.12
Beilstein:
741937
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39021208
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.5 (vs air)

vapor pressure

6 mmHg ( 20 °C)

Assay

≥99.6% (GC)

autoignition temp.

662 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

11.4 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.452 (lit.)
n20/D 1.452

bp

140.4 °C (lit.)

mp

−23 °C (lit.)

solubility

H2O: soluble

density

0.975 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)CC(C)=O

InChI

1S/C5H8O2/c1-4(6)3-5(2)7/h3H2,1-2H3

InChI key

YRKCREAYFQTBPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetylacetone is a β-diketone, which contains two carbonyl groups and one active methylene group.

Application

Acetylacetone has been used as a volatile eluent modifier to remediate the detrimental impacts of metal cations during the quantification of energy metabolites by ultrahigh pressure-liquid chromatography-electrospray-tandem mass spectrometry (UHPLC-ESI-MS). It has also been used as a derivatization agent for aldehydes.(3

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Automatized flow-batch method for fluorescent determination of free glycerol in biodiesel samples using on-line extraction
Lima MB, et al.
Talanta, 89(2), 21-26 (2012)
Controlling detrimental effects of metal cations in the quantification of energy metabolites via ultrahigh pressure-liquid chromatography-electrospray-tandem mass spectrometry by employing acetylacetone as a volatile eluent modifier
Siegel D, et al.
Journal of Chromatography A, 1294(2), 87-97 (2013)
2, 4-pentanedione
Ballantyne B
Journal of Applied Toxicology, 21(2), 165-171 (2001)
Liquid-phase microextraction with in-drop derivatization combined with microvolume fluorospectrometry for free and hydrolyzed formaldehyde determination in textile samples
Saenz M, et al.
Analytica Chimica Acta, 687(1), 50-55 (2011)
Giuliano Alagona et al.
Physical chemistry chemical physics : PCCP, 12(35), 10173-10188 (2010-08-03)
The catalytic effect of explicit water molecules on the keto-enol tautomerism in a system of biological interest (enolpyruvate) has been investigated at the B3LYP/6-31++G** level by exploring the potential energy surface in the presence of 1 or 2 water molecules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service