Merck
  • Characterization of Metabolic Patterns in Mouse Oocytes during Meiotic Maturation.
Characterization of Metabolic Patterns in Mouse Oocytes during Meiotic Maturation.
Molecular cell (2020-10-18)
Ling Li, Shuai Zhu, Wenjie Shu, Yueshuai Guo, Yusheng Guan, Juan Zeng, Haichao Wang, Longsen Han, Jiaqi Zhang, Xiaohui Liu, Chunling Li, Xiaojing Hou, Min Gao, Juan Ge, Chao Ren, Hao Zhang, Tim Schedl, Xuejiang Guo, Minjian Chen, Qiang Wang
ABSTRACT
Well-balanced and timed metabolism is essential for making a high-quality egg. However, the metabolic framework that supports oocyte development remains poorly understood. Here, we obtained the temporal metabolome profiles of mouse oocytes during in vivo maturation by isolating large number of cells at key stages. In parallel, quantitative proteomic analyses were conducted to bolster the metabolomic data, synergistically depicting the global metabolic patterns in oocytes. In particular, we discovered the metabolic features during meiotic maturation, such as the fall in polyunsaturated fatty acids (PUFAs) level and the active serine-glycine-one-carbon (SGOC) pathway. Using functional approaches, we further identified the key targets mediating the action of PUFA arachidonic acid (ARA) on meiotic maturation and demonstrated the control of epigenetic marks in maturing oocytes by SGOC network. Our data serve as a broad resource on the dynamics occurring in metabolome and proteome during oocyte maturation.
Sigma-Aldrich
Dihydroxyacetone phosphate hemimagnesium salt hydrate, ≥95% (TLC)
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Xanthosine dihydrate, ≥99%
Sigma-Aldrich
Inosine 5′-monophosphate from Saccharomyces cerevisiae, ≥98%
Sigma-Aldrich
Agmatine sulfate salt, ≥97%, powder
Sigma-Aldrich
all trans-Retinal, powder, ≥98%
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid, ≥98%
Sigma-Aldrich
L-Norvaline, arginase inhibitor
Sigma-Aldrich
Inosine, ≥99% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
L-(−)-3-Phenyllactic acid, 98%
Sigma-Aldrich
3-Phenylbutyric acid, 98%
Sigma-Aldrich
Tetradecanedioic acid, 99%
Sigma-Aldrich
D-Panthenol, ≥98.0% (NT)
Supelco
Pentadecanoic acid, analytical standard
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
BW A4C, ≥98% (HPLC)
Sigma-Aldrich
D-Saccharic acid potassium salt, ≥98% (GC)
Sigma-Aldrich
2′-Deoxycytidine, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Neopterin, ≥97.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Histamine dihydrochloride, ≥99.0% (AT)
Sigma-Aldrich
L-Tryptophanol, 97%
Sigma-Aldrich
Milrinone, ≥97% (TLC), powder
Sigma-Aldrich
N-(2-Aminoethyl)acetamide, technical grade, 90%
Sigma-Aldrich
Asp-Phe, 96%
Sigma-Aldrich
β-Alanine, 99%
Sigma-Aldrich
D-(−)-Quinic acid, 98%
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Anti-α-Tubulin−FITC antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture