Skip to Content
Merck

Skip To

D3897

2′-Deoxycytidine

≥99% (HPLC), synthetic (organic), powder

Synonym(s):

Cytosine deoxyriboside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H13N3O4
CAS Number:
Molecular Weight:
227.22
NACRES:
NA.51
PubChem Substance ID:
UNSPSC Code:
41106305
EC Number:
213-454-1
MDL number:
Beilstein/REAXYS Number:
87567
Assay:
≥99% (HPLC)
Biological source:
synthetic (organic)
Form:
powder
Solubility:
water: 50 mg/mL, clear, colorless to very faintly yellow
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

2′-Deoxycytidine, ≥99% (HPLC)

biological source

synthetic (organic)

Quality Level

assay

≥99% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

NC1=NC(=O)N(C=C1)[C@H]2C[C@H](O)[C@@H](CO)O2

InChI

1S/C9H13N3O4/c10-7-1-2-12(9(15)11-7)8-3-5(14)6(4-13)16-8/h1-2,5-6,8,13-14H,3-4H2,(H2,10,11,15)/t5-,6+,8+/m0/s1

InChI key

CKTSBUTUHBMZGZ-SHYZEUOFSA-N

General description

2′-Deoxycytidine (deoxyC) is one of the deoxynucleosides containing cytosine as the nucleobase. It is found in the blood, feces, and urine.

Application

2′-Deoxycytidine has been used:
  • as a substrate for Trypanosoma brucei cytidine deaminase (TbCDA) to measure its activity[1]
  • as a standard in the isolation and quantification of metabolite levels in murine tumor interstitial fluid by liquid chromatography-mass spectrometry (LC–MS)[2]
  • to study the role of autophagy in response to oncogenes and DNA replication stress[3]

Biochem/physiol Actions

2′-Deoxycytidine (deoxyC) forms dCTP upon phosphorylation which is used to synthesis DNA via various DNA polymerases or reverse transcriptases. DeoxyC is the substrate for deoxycytidine deaminase (EC 3.5.4.14) which converts it into 2′-deoxyuridine. DeoxyC is phosphorylated to the nucleotide dCMP by the enzyme deoxycytidine kinase (DCK). DeoxyC serves as a potential head and neck cancer marker.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D5782D5412D7400
biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

synthetic (organic)

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200

assay

≥99% (HPLC)

assay

≥98% (HPLC)

assay

≥98.5%

assay

≥99%

form

powder

form

powder

form

powder

form

powder

solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

solubility

water: 25 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Martyna Modrzejewska et al.
Free radical biology & medicine, 101, 378-383 (2016-11-12)
The most plausible mechanism behind active demethylation of 5-methylcytosine involves TET proteins which participate in oxidation of 5-methylcytosine to 5-hydroxymethylcytosine; the latter is further oxidized to 5-formylcytosine and 5-carboxycytosine. 5-Hydroxymethyluracil can be also generated from thymine in a TET-catalyzed process.
Urinary Nucleosides and Deoxynucleosides
Patejko M, et al.
Advances in Clinical Chemistry, 83, 1-51 (2018)
Megan Beetch et al.
Scientific reports, 10(1), 2415-2415 (2020-02-14)
Triple-negative breast cancer (TNBC) is a subtype of breast cancer unresponsive to traditional receptor-targeted treatments, leading to a disproportionate number of deaths. Invasive breast cancer is believed to evolve from non-invasive ductal carcinoma in situ (DCIS). Detection of triple-negative DCIS



Global Trade Item Number

SKUGTIN
D3897-500MG04061833047903
D3897-250MG04061833047897
D3897-100MG04061833047880
D3897-1G04061833563687
D3897-5G04061833047910

Questions

Reviews

No rating value

Active Filters