Skip to Content
Merck
All Photos(1)

Key Documents

N3386

Sigma-Aldrich

D-(+)-Neopterin

≥97.5% (sum of enantiomers, HPLC)

Synonym(s):

D-erythro-Neopterin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N5O4
CAS Number:
Molecular Weight:
253.21
Beilstein:
89070
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.5% (sum of enantiomers, HPLC)

form

powder

storage temp.

2-8°C

SMILES string

Nc1nc(O)c2nc(cnc2n1)[C@H](O)[C@H](O)CO

InChI

1S/C9H11N5O4/c10-9-13-7-5(8(18)14-9)12-3(1-11-7)6(17)4(16)2-15/h1,4,6,15-17H,2H2,(H3,10,11,13,14,18)/t4-,6+/m1/s1

InChI key

BMQYVXCPAOLZOK-XINAWCOVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(+)-Neopterin is a pteridine derivative, naturally found in Dictyostelium discoideum. It is found in significant amounts in the urine of cancer patients.

Application

D-(+)-Neopterin has been used as a NADPH oxidase inhibitor while assessing the ability of NADPH oxidases in breast cancer cells to produce reactive oxygen species (ROS). [NADPH= nicotinamide adenine dinucleotide phosphate]

Biochem/physiol Actions

Pterin cofactor; precursor of BH4 (tetrahydrobiopterin).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dictyopterin, 6-(d-threo-1,2-dihydroxypropyl)-pterin, a new natural isomer of L-biopterin.
Klein R, et al.
European Journal of Biochemistry, 187(3), 665-669 (1990)
Pteridine analysis in urine by capillary electrophoresis using laser-induced fluorescence detection.
Han F, et al.
Analytical Chemistry, 71(7), 1265-1269 (1999)
Ferroptosis is induced following siramesine and lapatinib treatment of breast cancer cells.
Ma S, et al.
Cell Death & Disease, 7(7), e2307-e2307 (2016)
Nisreen Husain et al.
Journal of clinical gastroenterology, 47(3), 246-251 (2012-12-28)
Neopterin, a pyrazino-[2,3-d]-pyrimidine compound, is a metabolite of cyclic guanosine monophosphate that is released by activated T lymphocytes and macrophages after induction by γ interferon. We sought to determine whether neopterin concentration in stool, serum, or urine is a useful
R Caruso et al.
Clinical biochemistry, 46(1-2), 94-98 (2012-10-30)
Neopterin, a marker of inflammation and monocyte activation, is found increased in patients with heart failure (HF). This study investigates whether neopterin levels correlate with left ventricular (LV) remodeling and brain natriuretic peptide (BNP), a marker of cardiac stress, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service