Merck
  • Genetically targeted chemical assembly of functional materials in living cells, tissues, and animals.
Genetically targeted chemical assembly of functional materials in living cells, tissues, and animals.
Science (New York, N.Y.) (2020-03-21)
Jia Liu, Yoon Seok Kim, Claire E Richardson, Ariane Tom, Charu Ramakrishnan, Fikri Birey, Toru Katsumata, Shucheng Chen, Cheng Wang, Xiao Wang, Lydia-Marie Joubert, Yuanwen Jiang, Huiliang Wang, Lief E Fenno, Jeffrey B-H Tok, Sergiu P Pașca, Kang Shen, Zhenan Bao, Karl Deisseroth
ABSTRACT
The structural and functional complexity of multicellular biological systems, such as the brain, are beyond the reach of human design or assembly capabilities. Cells in living organisms may be recruited to construct synthetic materials or structures if treated as anatomically defined compartments for specific chemistry, harnessing biology for the assembly of complex functional structures. By integrating engineered-enzyme targeting and polymer chemistry, we genetically instructed specific living neurons to guide chemical synthesis of electrically functional (conductive or insulating) polymers at the plasma membrane. Electrophysiological and behavioral analyses confirmed that rationally designed, genetically targeted assembly of functional polymers not only preserved neuronal viability but also achieved remodeling of membrane properties and modulated cell type-specific behaviors in freely moving animals. This approach may enable the creation of diverse, complex, and functional structures and materials within living systems.
Sigma-Aldrich
N-Phenyl-p-phenylenediamine, 98%
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ACS reagent, ≥98.5%
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride hydrate, ≥96%
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
Polyaniline (emeraldine base), average Mw ~50,000
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Buprenorphine, European Pharmacopoeia (EP) Reference Standard