Skip to Content
Merck
All Photos(1)

Key Documents

C127

Sigma-Aldrich

CNQX

≥98% (HPLC), solid

Synonym(s):

6-Cyano-7-nitroquinoxaline-2,3-dione, FG-9065

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4N4O4
CAS Number:
Molecular Weight:
232.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

yellow

solubility

DMSO: >10 mg/mL
H2O: insoluble

SMILES string

[O-][N+](=O)c1cc2NC(=O)C(=O)Nc2cc1C#N

InChI

1S/C9H4N4O4/c10-3-4-1-5-6(2-7(4)13(16)17)12-9(15)8(14)11-5/h1-2H,(H,11,14)(H,12,15)

InChI key

RPXVIAFEQBNEAX-UHFFFAOYSA-N

Related Categories

Application

CNQX has been used:
  • as an α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA)/kainate blocker to study its effects on cytoneme formation in pluripotent embryonic stem cells (pESCs)[1]
  • glutamate blocker to reduce the synaptic input during current-clamp recordings in central antennal lobe neurons[2]
  • as a non-N-methyl-D-aspartate (NMDA) receptor blocker to study its effects on the heart rate in rats[3]

Biochem/physiol Actions

Potent, competitive AMPA/kainate glutamate receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiyan Li et al.
Frontiers in cellular neuroscience, 11, 324-324 (2017-11-11)
Release of the major excitatory neurotransmitter glutamate by synaptic vesicle exocytosis depends on glutamate loading into synaptic vesicles by vesicular glutamate transporters (VGLUTs). The two principal isoforms, VGLUT1 and 2, exhibit a complementary pattern of expression in adult brain that
Guan Hock Khoo et al.
Molecular neurobiology, 56(9), 6436-6450 (2019-03-04)
Long-term depression (LTD) of synaptic efficacy is widely regarded as a cellular basis of learning and memory. The magnitude of hippocampal CA1 LTD induced by low-frequency stimulation (LFS) declines with age, but the mechanisms involved remain poorly understood. Perineuronal nets
Binbin Yan et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 296(2), R299-R308 (2008-11-21)
Chronic intermittent hypoxia (CIH), as occurs in sleep apnea, impairs baroreflex-mediated reductions in heart rate (HR) and enhances HR responses to electrical stimulation of vagal efferent. We tested the hypotheses that HR responses to activation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate (AMPA) and
Emmanuelle Berret et al.
Nature communications, 8(1), 557-557 (2017-09-17)
Oligodendrocyte (OL) maturation and axon-glial communication are required for proper myelination in the developing brain. However, physiological properties of OLs remain largely uncharacterized in different brain regions. The roles of oligodendroglial voltage-activated Na+ channels (Nav) and electrical excitability in relation
Ira Espuny-Camacho et al.
Neuron, 77(3), 440-456 (2013-02-12)
The study of human cortical development has major implications for brain evolution and diseases but has remained elusive due to paucity of experimental models. Here we found that human embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs), cultured

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service