Merck
  • Characterization, Dynamics, and Mechanism of CXCR4 Antagonists on a Constitutively Active Mutant.
Characterization, Dynamics, and Mechanism of CXCR4 Antagonists on a Constitutively Active Mutant.
Cell chemical biology (2019-03-05)
Eric M Rosenberg, Reed E S Harrison, Lun Kelvin Tsou, Natalie Drucker, Brock Humphries, Deepa Rajasekaran, Kathryn E Luker, Chien-Huang Wu, Jen-Shin Song, Chuan-Jen Wang, James W Murphy, Yung-Chi Cheng, Kak-Shan Shia, Gary D Luker, Dimitrios Morikis, Elias J Lolis
ABSTRACT
The G protein-coupled receptor (GPCR) CXCR4 is a co-receptor for HIV and is involved in cancers and autoimmune diseases. We characterized five purine or quinazoline core polyamine pharmacophores used for targeting CXCR4 dysregulation in diseases. All were neutral antagonists for wild-type CXCR4 and two were biased antagonists with effects on β-arrestin-2 only at high concentrations. These compounds displayed various activities for a constitutively active mutant (CAM). We use the IT1t-CXCR4 crystal structure and molecular dynamics (MD) simulations to develop two hypotheses for the activation of the N1193.35A CAM. The N1193.35A mutation facilitates increased coupling of TM helices III and VI. IT1t deactivates the CAM by disrupting the coupling between TM helices III and VI, mediated primarily by residue F872.53. Mutants of F872.53 in N1193.35A CXCR4 precluded constitutive signaling and prevented inverse agonism. This work characterizes CXCR4 ligands and provides a mechanism for N1193.35A constitutive activation.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial