Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium carbonate
Synonym(s):
Potassium carbonate
Linear Formula:
K
2
CO
3
CAS Number:
584-08-7
Molecular Weight:
138.21
Products
Product Category
salts (8)
inorganic bases (4)
Brand
Sigma-Aldrich (9)
Supelco (2)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (3)
Anhydrous (2)
Puriss (1)
Puriss P.A. (1)
pharmaceutical primary standard (1)
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
209619
Potassium carbonate,
ACS reagent, ≥99.0%
Expand
View Pricing
791776
Potassium carbonate,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99%
Expand
View Pricing
347825
Potassium carbonate,
reagent grade, ≥98%, powder, −325 mesh
Expand
View Pricing
60109
Potassium carbonate,
puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Expand
View Pricing
1.04928
Potassium carbonate,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
367877
Potassium carbonate,
99.995% trace metals basis
Expand
View Pricing
310263
Potassium carbonate,
ReagentPlus
®
, 99%
Expand
View Pricing
590681
Potassium carbonate,
anhydrous, powder, 99.99% trace metals basis
Expand
View Pricing
P5833
Potassium carbonate,
BioXtra, ≥99.0%
Expand
View Pricing
60108
Potassium carbonate,
BioUltra, anhydrous, ≥99.0% (T)
Expand
View Pricing
P1472
Potassium carbonate,
meets USP testing specifications
Expand
View Pricing
1548167
Potassium carbonate,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
1.10614
Potassium carbonate,
(special grade LAB)
Expand
View Pricing