Skip to Content
Merck
All Photos(3)

Documents

P5833

Sigma-Aldrich

Potassium carbonate

BioXtra, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

product line

BioXtra

Assay

≥99.0%

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

pH

11-13 (25 °C, 138 g/L)

mp

891 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.02%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.5%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium carbonate is an inorganic base that is used in the alkylation of phenols and 1,3-dicarbonyl compounds, as well as the generation of sulfur and phosphorus ylides. It is also used to dry organic solvents.

Application

Potassium carbonate can be used as a catalyst in the:
  • Markovnikov addition reactions.
  • Suzuki cross-coupling reactions.
It can also be used as a base for the N-arylation of benzamides via C-N coupling reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potassium carbonate as a green catalyst for Markovnikov addition of azoles to vinyl acetate in PEG
Kidwai M, et al.
Green Chemistry Letters and Reviews, 6, 63-68 (2013)
Application of experimental design and multivariate analysis in the on-line reaction monitoring of a Suzuki cross-coupling reaction by Raman spectroscopy and multivariate analysis
Hetemi D, et al.
Vibrational Spectroscopy, 100, 93-98 (2019)
Potassium Carbonate Promoted C-N Coupling Reaction between Benzamides and Aryl Iodides
Huang F, et al.
Synthesis, 50, 1090-1096 (2018)
Potassium Carbonate
Deshayes K
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Carlos Arroniz et al.
Organic letters, 15(4), 910-913 (2013-02-05)
ortho-Arylation of ortho-substituted benzoic acids is a challenging process due to the tendency of the reaction products toward Pd-catalyzed protodecarboxylation. A simple method for preventing decarboxylation in sterically hindered benzoic acids is reported. The method described represents a reliable and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service