Skip to Content
Merck
All Photos(3)

Key Documents

310263

Sigma-Aldrich

Potassium carbonate

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
solid
Pricing and availability is not currently available.

grade

reagent

product line

ReagentPlus®

Assay

99%

form

solid

pH

11-13 (25 °C, 138 g/L)

mp

891 °C (lit.)

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium carbonate is an inorganic base.[1] It participates as a base in the palladacycle catalyzed Heck reaction of chlorobenzene with styrene.[2] Addition of amino acids (glycine, sarcosine and proline) to K2CO3 (solvent) promotes the absorption of CO2.[3]

Application

Potassium carbonate may be used in the following processes:
  • As a reagent in the synthesis of polysubstituted iodobenzene derivatives.[4]
  • As a base for the Suzuki coupling of aryl halides with aryl boronic acids.[5]
  • The Heck reaction of styrene and bromobenzene.[1]
  • Preparation of disulfonated bis[4-(3-aminophenoxy)phenyl]sulfone, a disulfonated diamine monomer.[6]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A kinetic study of CO2 capture with potassium carbonate solutions promoted with various amino acids: glycine, sarcosine and proline.
Thee H, et al.
International Journal of Greenhouse Gas Control, 20, 212-222 (2014)
Chelating N-heterocyclic carbene ligands in palladium-catalyzed Heck-type reactions.
Herrmann WA, et al.
Journal of Organometallic Chemistry, 557(1), 93-96 (1998)
Robert Möckel et al.
Organic letters, 17(7), 1644-1647 (2015-03-21)
The cobalt-catalyzed Diels-Alder reaction of trimethylsilyl-substituted alkynes with 1,3-dienes led to dihydroaromatic intermediates which were transformed into iodobenzene derivatives. For this transformation, the dihydroaromatic intermediates had to be oxidized and the trimethylsilyl-substituted arene had to undergo a silicon-iodine exchange reaction.
Heck reaction using palladium complexed to dendrimers on silica.
Alper H, et al.
Canadian Journal of Chemistry, 78(6), 920-924 (2000)
Polyamide interfacial composite membranes prepared from m-phenylene diamine, trimesoyl chloride and a new disulfonated diamine.
Xie W, et al.
Journal of Membrane Science, 403, 152-161 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service