Skip to Content
Merck
All Photos(1)

Key Documents

1682206

USP

Triclosan

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Triclosan, 5-Chloro-2-(2,4-dichlorophenoxy)phenol, Irgasan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7Cl3O2
CAS Number:
Molecular Weight:
289.54
Beilstein:
2057142
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

triclosan

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1cc(Cl)ccc1Oc2ccc(Cl)cc2Cl

InChI

1S/C12H7Cl3O2/c13-7-1-3-11(9(15)5-7)17-12-4-2-8(14)6-10(12)16/h1-6,16H

InChI key

XEFQLINVKFYRCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

A component of cefsulodin-irgasan-novobiocin selection medium for Yersinia, a human pathogen that may contaminate animal-source food products.

Biochem/physiol Actions

It is an inhibitor of the enoyl-ACP (acyl-carrier protein) reductase component of type II fatty acid synthase (FAS-II) in bacteria and Plasmodium. It also inhibits mammalian fatty acid synthase (FASN), and may have anticarcinogenic activity.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leyre Lavilla Lerma et al.
Food microbiology, 44, 249-257 (2014-08-03)
Enterococcus faecalis and Enterococcus faecium isolated from various traditional fermented foods of both animal and vegetable origins have shown multidrug resistance to several antibiotics and tolerance to biocides. Reduced susceptibility was intra and inter-species dependent and was due to specific
M Périamé et al.
International journal of cosmetic science, 36(4), 386-395 (2014-05-16)
The aim of this study was to obtain a better understanding regarding the origin of recurrent contamination by Enterobacter gergoviae in diverse cosmetic formula. We studied 65 isolates collected from various sources (clinical, food, cosmetics). RAPD analysis using AP12H, REP
Nanhua Chen et al.
Antimicrobial agents and chemotherapy, 58(8), 4773-4781 (2014-06-11)
Artemisinin (ART)-based combination therapy (ACT) is used as the first-line treatment of uncomplicated falciparum malaria worldwide. However, despite high potency and rapid action, there is a high rate of recrudescence associated with ART monotherapy or ACT long before the recent
Lorillee C Tallorin et al.
Bioorganic & medicinal chemistry, 22(21), 6053-6061 (2014-10-07)
Enoyl-acyl carrier protein reductase (ENR), a critical enzyme in type II fatty acid biosynthesis, is a promising target for drug discovery against hepatocyte-stage Plasmodium falciparum. In order to identify PfENR-specific inhibitors, we docked 70 FDA-approved, bioactive, and/or natural product small
Likou Zou et al.
The Journal of antimicrobial chemotherapy, 69(10), 2644-2649 (2014-06-09)
To examine the distribution of all genes known to be responsible for resistance to quaternary ammonium compounds (QACs), and their association with resistance to QACs and other antimicrobials, in Escherichia coli recovered from retail meats. A total of 570 strains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service