Skip to Content
Merck
All Photos(1)

Key Documents

1642802

USP

Tacrolimus

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

FK-506 monohydrate, Tacrolimus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H69NO12 · H2O
CAS Number:
Molecular Weight:
822.03
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

tacrolimus

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.CO[C@@H]1C[C@@H](CC[C@H]1O)\C=C(/C)[C@H]2OC(=O)[C@@H]3CCCCN3C(=O)C(=O)[C@]4(O)O[C@H]([C@H](C[C@@H](C)C\C(C)=C\[C@@H](CC=C)C(=O)C[C@H](O)[C@H]2C)OC)[C@H](C[C@H]4C)OC

InChI

1S/C44H69NO12.H2O/c1-10-13-31-19-25(2)18-26(3)20-37(54-8)40-38(55-9)22-28(5)44(52,57-40)41(49)42(50)45-17-12-11-14-32(45)43(51)56-39(29(6)34(47)24-35(31)48)27(4)21-30-15-16-33(46)36(23-30)53-7;/h10,19,21,26,28-34,36-40,46-47,52H,1,11-18,20,22-24H2,2-9H3;1H2/b25-19+,27-21+;/t26-,28+,29+,30-,31+,32-,33+,34-,36+,37-,38-,39+,40+,44+;/m0./s1

InChI key

NWJQLQGQZSIBAF-MLAUYUEBSA-N

Gene Information

human ... FKBP1A(2280)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tacrolimus USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Tacrolimus Capsules
  • Tacrolimus Compounded Oral Suspension

Biochem/physiol Actions

FK-506 is a potent immunosuppressant, neuroprotective and neuroregenerative, and in vitro T cell proliferation blocker. FK-506 disrupts calcineurin-mediated signal transduction in T lymphocytes and interacts with its FK-506-binding protein-12 (FKBP12).
FK-506 is a potent immunosuppressant; neuroprotective and neuroregenerative; in vitro T cell proliferation blocker.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tacrolimus Compounded Oral Suspension
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(1), 4202-4202 (2016)
Tacrolimus capsules
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(2), 4196-4196 (2022)
Tacrolimus
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(2), 4193-4193 (2021)
Frank Stifft et al.
Transplantation, 97(7), 775-780 (2014-04-02)
Tacrolimus has originally been registered as a twice-daily formulation (Prograf, Tac BID), although a once-daily formulation (Advagraf, Tac QD) is also available. A reduced intrapatient variability of Tac Cmin, a surrogate marker for 24-hour drug exposure (AUC0-24), has been suggested.
Violette M G J Gijsen et al.
Pediatric transplantation, 17(3), 205-215 (2013-03-02)
Renal dysfunction after non-renal transplantation in adult tacrolimus-treated transplant patients is well documented. Little is known about its prevalence in children. Age-related changes in both disposition and effect of tacrolimus as well as renal function may preclude extrapolation of adult

Questions

  1. How do I reconstitute this product (100mg), and what is the recommended dosage to treat 1x10^6 cells (human PBMC) with this product?

    1 answer
    1. This product is freely soluble in ethanol. No recommended dosage for cells is available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service