Skip to Content
Merck
All Photos(1)

Key Documents

1604654

USP

Risperidone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

R 64,766

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27FN4O2
CAS Number:
Molecular Weight:
410.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

risperidone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC1=C(CCN2CCC(CC2)c3noc4cc(F)ccc34)C(=O)N5CCCCC5=N1

InChI

1S/C23H27FN4O2/c1-15-18(23(29)28-10-3-2-4-21(28)25-15)9-13-27-11-7-16(8-12-27)22-19-6-5-17(24)14-20(19)30-26-22/h5-6,14,16H,2-4,7-13H2,1H3

InChI key

RAPZEAPATHNIPO-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Risperidone is a new central 5-hydroxytryptamine2 and dopamine D2 antagonist. It is used to treat agitation, aggression,and psychosis caused during demnetia. It is a neuroleptic drug. It is also an atypical antipsychotic drug which is used for treating schizophrenia.

Application

It was used as reference standard during RP-UPLC analysis for determination of risperidone related compounds in Risperdal® Consta (risperidone long-acting injection).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Legal Information

Risperdal is a registered trademark of Johnson & Johnson

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and Validation of RP-UPLC Method for Determination of Related Substances in Risperdal? Consta?.
Chen, Cheng-Tung, Cheng-Wei Cheng, and Yu-Fang Hu.
Journal of Chromatography and Separation Techniques, 6.6, 1-1 (2015)
Jérôme A J Becker et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(9), 2049-2060 (2014-03-13)
The etiology of Autism Spectrum Disorders (ASDs) remains largely unknown. Identifying vulnerability genes for autism represents a major challenge in the field and allows the development of animal models for translational research. Mice lacking the mu opioid receptor gene (Oprm1(-/-))
Christian Spang Pedersen et al.
Behavioural brain research, 273, 63-72 (2014-07-30)
Schizophrenia is a severe psychiatric disorder characterized by three symptom domains, positive (hallucinations, obsession), negative (social withdrawal, apathy, self-neglect) and cognitive (impairment in attention, memory and executive function). Whereas current medication ameliorates positive symptomatology, negative symptoms as well as cognitive
A comparison of risperidone and haloperidol for the prevention of relapse in patients with schizophrenia.
Csernansky JG
The New England Journal of Medicine, 346(1), 16-22 (2002)
Katherine Seto et al.
Therapeutic drug monitoring, 33(3), 275-283 (2011-03-26)
Although risperidone is commonly used in the acute and maintenance treatment of schizophrenia, the role of therapeutic drug monitoring has yet to be elucidated. The purpose of this review was to determine whether risperidone warrants therapeutic drug monitoring in patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service