Skip to Content
Merck
All Photos(1)

Key Documents

R3030

Sigma-Aldrich

Risperidone

≥98% (HPLC), powder, antipsychotic

Synonym(s):

R 64,766

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27FN4O2
CAS Number:
Molecular Weight:
410.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Risperidone, ≥98% (HPLC), powder

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥5 mg/mL

originator

Johnson & Johnson

storage temp.

room temp

SMILES string

CC1=C(CCN2CCC(CC2)c3noc4cc(F)ccc34)C(=O)N5CCCCC5=N1

InChI

1S/C23H27FN4O2/c1-15-18(23(29)28-10-3-2-4-21(28)25-15)9-13-27-11-7-16(8-12-27)22-19-6-5-17(24)14-20(19)30-26-22/h5-6,14,16H,2-4,7-13H2,1H3

InChI key

RAPZEAPATHNIPO-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Risperidone is a benzisoxazole derivative. It functions as an antagonist for dopamine D2, histamine H1 and α1 and α2 adrenergic receptors. Risperidone is used to treat schizophrenia, bipolar disorder and autism.

Application

Risperidone has been used:
  • to study its effects on bone formation and differentiation
  • to investigate the relationship between risperidone (RIS) dosages and RIS plasma levels in autism spectrum disorder (ASD) pediatric patients
  • to reverse induced schizophrenia-like behavior in mice

Biochem/physiol Actions

Risperidone is an antipsychotic; serotonin-dopamine antagonist.

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of liquid chromatography/tandem mass spectrometry analysis for therapeutic drug monitoring of risperidone and 9-hydroxyrisperidone in pediatric patients with autism spectrum disorders
Vanwong N, et al.
Journal of Clinical Laboratory Analysis, 30(6), 1236-1246 (2016)
Schizophrenia (2006)
Overexpression of transmembrane protein 168 in the mouse nucleus accumbens induces anxiety and sensorimotor gating deficit
Fu K, et al.
PLoS ONE, 12(12), e0189006-e0189006 (2017)
Kaplan & Sadock's Concise Textbook of Clinical Psychiatry (2008)
Atsumi Nitta et al.
Journal of personalized medicine, 11(7) (2021-07-03)
Piccolo, a presynaptic cytomatrix protein, plays a role in synaptic vesicle trafficking in the presynaptic active zone. Certain single-nucleotide polymorphisms of the Piccolo-encoding gene PCLO are reported to be associated with mental disorders. However, a few studies have evaluated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service