Skip to Content
Merck
All Photos(1)

Documents

1483152

USP

Oxcarbazepine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

10,11-Dihydro-10-oxo-5h-dibenz[b,f]azepine-5-carboxamide, Oxacarbazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxcarbazepine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C1CC2=C(C=CC=C2)N(C(N)=O)C3=CC=CC=C31

InChI

1S/C15H12N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8H,9H2,(H2,16,19)

InChI key

CTRLABGOLIVAIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxcarbazepine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Oxcarbazepine Oral Suspension
  • Oxcarbazepine Tablets

Biochem/physiol Actions

Anticonvulsant, antineuralgic. Inhibits veratrine-induced transmitter release.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara R Sommer et al.
Expert opinion on drug safety, 6(2), 133-145 (2007-03-21)
Few controlled studies are available to guide the clinician in treating potentially assaultive elderly individuals with psychiatric disorders. Safety concerns limit the use of benzodiazepines and antipsychotic medications in the elderly individual, making anticonvulsants an attractive alternative. This paper reviews
A Vasudev et al.
The Cochrane database of systematic reviews, (1)(1), CD005171-CD005171 (2008-02-07)
Some studies have suggested that oxcarbazepine has a role in preventing episode recurrence in bipolar affective disorder. This review attempted to investigate the existing evidence from randomised controlled trials for its use in the maintenance treatment of this illness. To
Gaetano Zaccara et al.
Seizure, 22(7), 528-536 (2013-04-30)
Analysis of overall tolerability and neurological adverse effects (AEs) of eslicarbazepine acetate (ESL), lacosamide (LCM) and oxcarbazepine (OXC) from double-blind, placebo-controlled trials. Indirect comparisons of patients withdrawing because of AEs, and the incidence of some vestibulocerebellar AEs between these three
Wetid Pratoomsri et al.
Canadian journal of psychiatry. Revue canadienne de psychiatrie, 51(8), 540-545 (2006-08-29)
To review the data on the efficacy of oxcarbazepine (OXC) in bipolar disorder (BD) and to provide recommendations for clinicians on the use of this medication in treating BD. Using the terms oxcarbazepine and bipolar disorder, oxcarbazepine and mania, or
Penny Bring et al.
Clinical pharmacokinetics, 47(12), 767-778 (2008-11-26)
Oxcarbazepine, the 10-keto analogue of carbamazepine, is approved for the treatment of partial seizures or generalized tonic-clonic seizures. The primary metabolite of oxcarbazepine is monohydroxylated derivative (MHD). This review follows a decision-making algorithm to determine if therapeutic drug monitoring of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service