Skip to Content
Merck

Skip To

1478367

USP

Olmesartan medoxomil

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C29H30N6O6
CAS Number:
Molecular Weight:
558.59
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

olmesartan

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCC1=NC(C(O)(C)C)=C(C(OCC2=C(C)OC(O2)=O)=O)N1CC(C=C3)=CC=C3C4=CC=CC=C4C5=NN=NN5

InChI

1S/C29H30N6O6/c1-5-8-23-30-25(29(3,4)38)24(27(36)39-16-22-17(2)40-28(37)41-22)35(23)15-18-11-13-19(14-12-18)20-9-6-7-10-21(20)26-31-33-34-32-26/h6-7,9-14,38H,5,8,15-16H2,1-4H3,(H,31,32,33,34)

InChI key

UQGKUQLKSCSZGY-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Olmesartan medoxomil USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0001405SML1391BP1173
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

BP

format

neat

format

neat

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical
pharmaceutical small molecule

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Gene Information

human ... AGTR1(185)

Gene Information

human ... AGTR1(185)

Gene Information

human ... AGTR1(185)

Gene Information

human ... AGTR1(185)


Still not finding the right product?

Explore all of our products under Olmesartan medoxomil


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1478367-100MG04061838737618

Questions

Reviews

No rating value

Active Filters