Skip to Content
Merck
All Photos(1)

Key Documents

1375105

USP

Mannitol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-Mannitol, ((2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol), 1,2,3,4,5,6-Hexanehexol, Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

mannitol

manufacturer/tradename

USP

mp

167-170 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mannitol USP reference standard for use in specified compendial tests and assays.

Also used to prepare standard preparation, resolution, system suitability, and standard solution for assay, identification, and impurity analysis according to the given below monographs of United States Pharmacopeia (USP):

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xylitol
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(4), 6105-6105 (2020)
Mannitol Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 28(1), 2736-2736 (2018)
Inositol
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 33(4), 5829-5829 (2020)
Mannitol
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(5), 2734-2734 (2020)
Isomalt
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 32(4), 5836-5836 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service