Skip to Content
Merck
All Photos(1)

Key Documents

1134324

USP

Ciprofloxacin Ethylenediamine Analog

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

7-[(2-Aminoethyl)amino]-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16FN3O3 · HCl
CAS Number:
Molecular Weight:
341.77
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ciprofloxacin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Fc1cc2c(cc1NCC[N+H3])N(C=C(C2=O)C(=O)O)C3CC3.[Cl-]

InChI

1S/C15H16FN3O3.ClH/c16-11-5-9-13(6-12(11)18-4-3-17)19(8-1-2-8)7-10(14(9)20)15(21)22;/h5-8,18H,1-4,17H2,(H,21,22);1H

InChI key

ODOFTUIHVVTJRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ciprofloxacin Ethylenediamine Analog USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ciprofloxacin Injection
  • Ciprofloxacin
  • Ciprofloxacin Tablets
  • Ciprofloxacin Ophthalmic Solution
  • Ciprofloxacin Ophthalmic Ointment
  • Ciprofloxacin and Dexamethasone Otic Suspension
  • Ciprofloxacin Extended-Release Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ciprofloxacin Extended-Release Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 37(6), 1017-1017 (2015)
Ciprofloxacin Hydrochloride
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(5), 1011-1011 (2020)
Ciprofloxacin Ophthalmic Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(3) (2020)
Ciprofloxacin Ophthalmic Ointment
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 45(6) (2021)
Ciprofloxacin Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 35(4), 1012-1012 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service