Skip to Content
Merck
All Photos(1)

Key Documents

1361009

USP

Levodopa

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3,4-Dihydroxy-L-phenylalanine, 3-(3,4-Dihydroxyphenyl)-L-alanine, L-3-Hydroxytyrosine, L-DOPA, Levodopa

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
197.19
Beilstein:
2215169
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levodopa

manufacturer/tradename

USP

mp

276-278 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N[C@@H](Cc1ccc(O)c(O)c1)C(O)=O

InChI

1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)/t6-/m0/s1

InChI key

WTDRDQBEARUVNC-LURJTMIESA-N

Gene Information

human ... DRD3(1814)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levodopa USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Carbidopa and Levodopa Tablets
  • Carbidopa and Levodopa Orally Disintegrating Tablets
  • Carbidopa and Levodopa Extended-Release Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Levodopa
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(1), 2604-2604 (2020)
Carbidopa and Levodopa Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 47(5) (2022)
Carbidopa and Levodopa Orally Disintegrating Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(1), 759-759 (2022)
Carbidopa and Levodopa Extended-Release Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(3), 753-753 (2018)
Vincent J J Odekerken et al.
Neurology, 84(13), 1355-1361 (2015-03-01)
To assess the neuropsychological outcome 12 months after bilateral deep brain stimulation (DBS) of the globus pallidus pars interna (GPi) or subthalamic nucleus (STN) for advanced Parkinson disease. We randomly assigned patients to receive either GPi DBS or STN DBS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service