Skip to Content
Merck
All Photos(1)

Key Documents

1286005

USP

Folic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

PteGlu, Pteroyl-L-glutamic acid, Vitamin M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19N7O6
CAS Number:
Molecular Weight:
441.40
Beilstein:
100781
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

folic acid

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NC(N1)=NC(C2=C1N=CC(CNC3=CC=C(C(N[C@@H](CCC(O)=O)C(O)=O)=O)C=C3)=N2)=O

InChI

1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1

InChI key

OVBPIULPVIDEAO-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Folic acid USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Folic Acid Tablets
  • Folic Acid Injection
  • Folic Acid Compounded Oral Solution
  • Oil- and Water-Soluble Vitamins with Minerals Chewable Gels
  • Leucovorin Calcium
  • Calcium l-5-Methyltetrahydrofolate Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Folic Acid Compounded Oral Solution
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(5), 2027-2027 (2017)
Folic Acid assay
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 48(3), 6689-6689 (2022)
Folic Acid
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 29(2), 2026-2026 (2020)
Folic Acid Tablets
United States Pharmacopeia, 48(3), 2028-2028 (2022)
Myra A Carpenter et al.
Journal of the American Society of Nephrology : JASN, 25(7), 1554-1562 (2014-03-15)
The optimal BP level in kidney transplant recipients remains uncertain. This post hoc analysis of the Folic Acid for Vascular Outcome Reduction in Transplantation (FAVORIT) trial cohort assessed associations of BP with a pooled cardiovascular disease (CVD) outcome and with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service