Skip to Content
Merck
All Photos(1)

Key Documents

1087009

USP

Calcium Pantothenate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-Pantothenic acid hemicalcium salt, (R)-(+)-N-(2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl)-β-alanine hemicalcium salt, Calcium D-pantothenate, Vitamin B5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C(CH3)2CH(OH)CONHCH2CH2CO2 ·1/2Ca
CAS Number:
Molecular Weight:
238.27
Beilstein:
3769272
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

calcium pantothenate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)O[Ca]OC(=O)CCNC(=O)[C@H](O)C(C)(C)CO

InChI

1S/2C9H17NO5.Ca/c2*1-9(2,5-11)7(14)8(15)10-4-3-6(12)13;/h2*7,11,14H,3-5H2,1-2H3,(H,10,15)(H,12,13);/q;;+2/p-2/t2*7-;/m00./s1

InChI key

FAPWYRCQGJNNSJ-UBKPKTQASA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Calcium Pantothenate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Calcium Pantothenate Tablets
  • Oil- and Water-Soluble Vitamins Capsules
  • Oil- and Water-Soluble Vitamins Oral Solution
  • Oil- and Water-Soluble Vitamins Tablets
  • Oil- and Water-Soluble Vitamins with Minerals Capsules
  • Oil- and Water-Soluble Vitamins with Minerals Oral Solution
Precursor in the biosynthesis of coenzyme A.

Physical properties

Due to the unstable, hygroscopic nature of the free acid, the calcium salt is employed.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maha R Farhat et al.
Genome medicine, 6(11), 101-101 (2014-12-09)
Whole genome sequencing is increasingly used to study phenotypic variation among infectious pathogens and to evaluate their relative transmissibility, virulence, and immunogenicity. To date, relatively little has been published on how and how many pathogen strains should be selected for
Daniel R G Price et al.
BMC biology, 12, 110-110 (2014-12-21)
Genome evolution in intracellular microbial symbionts is characterized by gene loss, generating some of the smallest and most gene-poor genomes known. As a result of gene loss these genomes commonly contain metabolic pathways that are fragmented relative to their free-living
Henning Frerigmann et al.
Plant physiology, 166(1), 349-369 (2014-07-23)
By means of yeast (Saccharomyces cerevisiae) two-hybrid screening, we identified basic helix-loop-helix transcription factor05 (bHLH05) as an interacting partner of MYB51, the key regulator of indolic glucosinolates (GSLs) in Arabidopsis (Arabidopsis thaliana). Furthermore, we show that bHLH04, bHLH05, and bHLH06/MYC2
Katsumi Shibata et al.
Bioscience, biotechnology, and biochemistry, 78(12), 2102-2109 (2014-08-15)
We previously reported that mild food restriction induces a reduction in tryptophan-nicotinamide conversion, which helps to explain why death secondary to pellagra is pandemic during the hungry season. In this study, we investigated the levels of B-group vitamins in the
Nathaniel W Snyder et al.
Rapid communications in mass spectrometry : RCM, 28(16), 1840-1848 (2015-01-07)
Acyl-Coenzyme A (CoA) thioesters are the principal form of activated carboxylates in cells and tissues. They are employed as acyl carriers that facilitate the transfer of acyl groups to lipids and proteins. Quantification of medium- and long-chain acyl-CoAs represents a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service