Skip to Content
Merck

Skip To

1179504

USP

Dexpanthenol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-Panthenol, (R)-(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, (R)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide, D-Pantothenyl alcohol, Dexpanthenol, Provitamin B

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H19NO4
CAS Number:
Molecular Weight:
205.25
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1724947
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

dexpanthenol

manufacturer/tradename

USP

density

1.20 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCCO

InChI

1S/C9H19NO4/c1-9(2,6-12)7(13)8(14)10-4-3-5-11/h7,11-13H,3-6H2,1-2H3,(H,10,14)/t7-/m0/s1

InChI key

SNPLKNRPJHDVJA-ZETCQYMHSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dexpanthenol USP reference standard for use in specified quality tests and assays.

Also used to prepare standard, and standard stock solutions for assay and strength analysis according to the given below general chapter and monographs of United States Pharmacopeia (USP):
  • 〈115〉 Dexpanthenol assay
  • Oil- and Water-Soluble Vitamins Oral Solution
  • Water-Soluble Vitamins with Minerals Oral Solution
  • Oil- and Water-Soluble Vitamins with Minerals Oral Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D0730000PHR122876200
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

application(s)

-

format

neat

format

neat

format

neat

format

-

density

1.20 g/mL at 20 °C (lit.)

density

1.20 g/mL at 20 °C (lit.)

density

1.20 g/mL at 20 °C (lit.)

density

1.20 g/mL at 20 °C (lit.)

API family

dexpanthenol

API family

dexpanthenol

API family

dexpanthenol

API family

-


Still not finding the right product?

Explore all of our products under Dexpanthenol


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Oil- and Water-Soluble Vitamins Oral Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(5), 5409-5409 (2016)
Oil- and Water-Soluble Vitamins with Minerals Oral Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(5), 5462-5462 (2016)
Water-Soluble Vitamins with Minerals Oral Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(5), 5543-5543 (2015)



Global Trade Item Number

SKUGTIN
1179504-500MG04061838705525

Questions

Reviews

No rating value

Active Filters