Skip to Content
Merck
All Photos(1)

Key Documents

1173100

USP

Desmethoxycurcumin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-(4-Hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O5
CAS Number:
Molecular Weight:
338.35
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

curcumin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O(C)c1c(ccc(c1)\C=C\C(=O)CC(=O)\C=C\c2ccc(cc2)O)O

InChI

1S/C20H18O5/c1-25-20-12-15(6-11-19(20)24)5-10-18(23)13-17(22)9-4-14-2-7-16(21)8-3-14/h2-12,21,24H,13H2,1H3/b9-4+,10-5+

InChI key

HJTVQHVGMGKONQ-LUZURFALSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Desmethoxycurcumin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Curcuminoids Tablets
  • Curcuminoids
  • Powdered Turmeric Extract
  • Powdered Turmeric
  • Turmeric

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Lan Jiang et al.
Chemical biology & drug design, 81(5), 650-657 (2013-01-29)
Traditionally, active compounds were discovered from natural product extracts by bioassay-guided fractionation, which was with high cost and low efficiency. A well-trained support vector regression model based on mean impact value was used to identify lead active compounds on inhibiting
Ming-Jyh Sheu et al.
Molecular nutrition & food research, 57(9), 1586-1597 (2013-03-23)
Curcumin has been shown to affect platelet-derived growth factor (PDGF)- and tumor necrosis factor (TNF)-α-elicited vascular smooth muscle cell (VSMC) migration and inhibit neointima formation following vascular injury. However, whether two other curcuminoids isolated from Curcuma longa, demethoxycurcumin (DMC) and
Yanyu Xiao et al.
Journal of agricultural and food chemistry, 61(15), 3654-3660 (2013-03-05)
Curcuminoid, a dietary polyphenolic compound, has poor water solubility and low bioavailability following oral administration. The aim of this study was to develop a formulation of curcuminoid-loaded microemulsion (Cur-ME) to improve its oral bioavailability. The optimized Cur-ME formulation was prepared
Yan Shen et al.
Journal of pharmaceutical and biomedical analysis, 81-82, 146-150 (2013-05-08)
A sensitive and efficient method based on pressurized liquid extraction (PLE) followed by liquid chromatography-tandem mass spectrometry (LC-MS/MS) was developed for simultaneous identification and quantification of three Curcuminoids in Curcuma wenyujin Y.H.chen et C.Ling., a well known traditional Chinese medicinal
Oliver B Villaflores et al.
Taiwanese journal of obstetrics & gynecology, 51(4), 515-525 (2013-01-02)
Alzheimer disease (AD) is by far the most common cause of dementia globally. This neurodegenerative disorder of the brain is chronic and progressive, characterized clinically by the deterioration in the key symptoms of behavioral and cognitive abilities. Treatment options for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service