Skip to Content
Merck
All Photos(1)

Documents

1118394

USP

Chloroquine Related Compound G

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-((7-Chloroquinolin-4-yl)amino)-N,N-diethylpentan-1-amine oxide sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26ClN3O · H2SO4
Molecular Weight:
433.95
UNSPSC Code:
41116107
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

USP

API family

chloroquine

form

powder

packaging

pkg of 15 mg

manufacturer/tradename

USP

storage condition

protect from light

color

yellow

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

Chloroquine Related Compound G is an impurity of chloroquine, which is a 9-aminoquinoline drug that effectively inhibits viral infections in humans.

Application

Chloroquine Related Compound G USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Chloroquine Phosphate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chloroquine is a potent inhibitor of SARS coronavirus infection and spread
Vincent MJ, et al.
Virology Journal, 2(1), 1-10 (2005)
Chloroquine Phosphate USP Monograph
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(3), 959-959 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service