Skip to Content
Merck
All Photos(1)

Key Documents

1118350

USP

Chloroquine Related Compound D

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

7-Chloro-4-{[4-(ethylamino)-1-methylbutyl]amino}quinoline, Monodesethylchloroqu​ine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22ClN3
CAS Number:
Molecular Weight:
291.82
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

USP

API family

chloroquine

form

powder

packaging

pkg of 15 mg

manufacturer/tradename

USP

storage condition

protect from light

color

tan

bp

343.4-347 °F (173—175°C; 845.06°F )

mp

176-215.6 °F (80—102°C)

solubility

chloroform: soluble
dichloromethane: soluble

density

1.14 g/cm3

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC2=C1C(NC(CCCNCC)C)=CC=N2

InChI

1S/C16H22ClN3/c1-3-18-9-4-5-12(2)20-15-8-10-19-16-11-13(17)6-7-14(15)16/h6-8,10-12,18H,3-5,9H2,1-2H3,(H,19,20)

InChI key

MCYUUUTUAAGOOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroquine Related Compound D is an impurity of chloroquine, which is a 9-aminoquinoline drug that effectively inhibits viral infections in humans.

Application

Chloroquine Related Compound D USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Chloroquine Phosphate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC with ultraviolet detection for the determination of chloroquine and desethylchloroquine in whole blood and finger-prick capillary blood dried on filter paper
Cheomung A and Na-Bangchang K
Journal of Pharmaceutical and Biomedical Analysis, 55(5), 1031-1040 (2011)
An HPLC method with diode array detector for the simultaneous quantification of chloroquine and desethylchloroquine in plasma and whole blood samples from Plasmodium vivax patients in Vietnam, using quinine as an internal standard
Van Pham T, et al.
Biomedical Chromatography, 30(7), 1104-1111 (2016)
Chloroquine Phosphate USP Monograph
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(3), 959-959 (2020)
Chloroquine is a potent inhibitor of SARS coronavirus infection and spread
Vincent MJ, et al.
Virology Journal, 2(1), 1-10 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service