Skip to Content
Merck
All Photos(1)

Key Documents

1103003

USP

Cetyl alcohol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Hexadecanol, Cetyl alcohol, Palmityl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15OH
CAS Number:
Molecular Weight:
242.44
Beilstein:
1748475
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

vapor density

8.34 (vs air)

vapor pressure

<0.01 mmHg ( 43 °C)

API family

cetyl alcohol

autoignition temp.

483 °F

expl. lim.

8 %

manufacturer/tradename

USP

bp

179-181 °C/10 mmHg (lit.)

mp

48-50 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCCCCCCO

InChI

1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

BXWNKGSJHAJOGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cetyl alcohol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Cetyl Esters Wax
  • Cetostearyl Alcohol
  • Hydrogenated Lanolin
  • Sodium Cetostearyl Sulfate
  • Oleyl Alcohol
  • Stearyl Alcohol

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cetyl Esters Wax
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(5), 5706-5706 (2018)
Cetostearyl Alcohol
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 48(4), 5702-5702 (2015)
Cetyl alcohol
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(3), 5705-5705 (2020)
Yoshinori Onuki et al.
Drug development and industrial pharmacy, 40(7), 937-943 (2013-05-02)
A non-destructive method for monitoring creaming of emulsion-based formulations is in great demand because it allows us to understand fully their instability mechanisms. This study was aimed at demonstrating the usefulness of magnetic resonance (MR) techniques, including MR imaging (MRI)
Mahmoud Mokhtar Ibrahim et al.
Drug development and industrial pharmacy, 40(6), 765-773 (2013-04-11)
In this study, unilamellar liposomal vesicles of diltiazem HCl (DH) were prepared using either reversed phase evaporation (REV) or proliposome methods. Soya phosphatidylcholine (SPC) was used for preparing the liposomes, and the vesicles were rigidified using cholesterol (Chol) or cetyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service